EMAIL THIS PAGE TO A FRIEND
Oncotarget
CXCL16/CXCR6 chemokine signaling mediates breast cancer progression by pERK1/2-dependent mechanisms.
PMID 25909173
Abstract
Our previous studies demonstrate that CXCL6/CXCR6 chemokine axis induces prostate cancer progression by the AKT/mTOR signaling pathway; however, its role and mechanisms underlying invasiveness and metastasis of breast cancer are yet to be elucidated. In this investigation, CXCR6 protein expression was examined using high-density tissue microarrays and immunohistochemistry. Expression of CXCR6 shows a higher epithelial staining in breast cancer nest site and metastatic lymph node than the normal breast tissue, suggesting that CXCR6 may be involved in breast cancer (BC) development. In vitro and in vivo experiments indicate that overexpression of CXCR6 in BC cells has a marked effect on increasing cell migration, invasion and metastasis. In contrast, reduction of CXCR6 expression by shRNAs in these cells greatly reduce its invasion and metastasis ability. Mechanistic analyses show that CXCL16/CXCR6 chemokine axis is capable of modulating activation of RhoA through activating ERK1/2 signaling pathway, which then inhibits the activity of cofilin, thereby enhancing the stability of F-actin, responsible for invasiveness and metastasis of BC. Taken together, our data shows for the first time that the CXCR6 / ERK1/2/ RhoA / cofilin /F-actin pathway plays a central role in the development of BC. Targeting the signaling pathway may prove beneficial to prevent metastasis and provide a more effective therapeutic strategy for BC.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2501321
Anti-CXCR6 (C-terminal) antibody produced in goat, affinity isolated antibody, buffered aqueous solution
AV07030
Anti-CXCR6 antibody produced in rabbit, affinity isolated antibody
HPA047591
Anti-CXCR6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410119
Anti-CXCR6 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4300214
Anti-phospho-RPS6KB1 (pSer411) antibody produced in rabbit, affinity isolated antibody
SAB4300225
Anti-phospho-RPS6KB1 (pSer424) antibody produced in rabbit, affinity isolated antibody
SAB4300202
Anti-phospho-RPS6KB1 (pThr421) antibody produced in rabbit, affinity isolated antibody
SAB4300444
Anti-RPS6KB1 (Ab-389) antibody produced in rabbit, affinity isolated antibody
SAB4300515
Anti-RPS6KB1 (Ab-411) antibody produced in rabbit, affinity isolated antibody
SAB4300500
Anti-RPS6KB1 (Ab-421) antibody produced in rabbit, affinity isolated antibody
SAB4300527
Anti-RPS6KB1 (Ab-424) antibody produced in rabbit, affinity isolated antibody
SAB1412617
ANTI-RPS6KB1 antibody produced in mouse, clone 2C2, purified immunoglobulin, buffered aqueous solution
SAB1412626
ANTI-RPS6KB1 antibody produced in mouse, clone 4H4, purified immunoglobulin, buffered aqueous solution
SAB2104784
Anti-RPS6KB1, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB2104785
Anti-RPS6KB1, (N-terminal) antibody produced in rabbit, affinity isolated antibody
S4047 Anti-S6 Kinase antibody produced in rabbit, whole antiserum
SAB4502691
Anti-S6K antibody produced in rabbit, affinity isolated antibody
SAB1404335
Monoclonal Anti-RPS6KB1, (C-terminal) antibody produced in mouse, clone 1E10, purified immunoglobulin, buffered aqueous solution