EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
Low-temperature phase transformation studies in the stearic acid: C form.
PMID 25909902
Abstract
This paper reports the temperature-dependent measurements in the C form of stearic acid. Raman scattering, X-ray diffraction, and differential scanning calorimetry measurements were performed at low temperatures. The polarized Raman spectra were measured for temperatures ranging from 8 to 300 K over the spectral range of 30-3000 cm(-1). The spectral changes observed in both the lattice vibrational modes and the internal vibrational modes regions of the Raman spectrum, allowed to identify a phase transition undergone by the stearic acid crystal occurring between 210 and 170 K and a change in the structure continues to be observed down to 8 K. The anharmonicity of some vibrational modes and the possible space groups presented by the crystal at low temperatures were also discussed. Low-temperature X-ray diffraction measurements were performed from 290 to 80 K and the results showed slight changes in the lattice parameters at ∼200 K. Furthermore, the evidence of the phase transformation was provided by the differential scanning calorimetry measurements, which identified an enthalpic anomaly at about 160 K.