EMAIL THIS PAGE TO A FRIEND
Endocrinology
A Nonradioactive Uptake Assay for Rapid Analysis of Thyroid Hormone Transporter Function.
PMID 25910050
Abstract
Thyroid hormones (TH) are actively taken up into target cells via TH-transmembrane transporters (THTT). Their activity and expression patterns define a layer of endocrine regulation that is poorly understood. Therefore, THTT are potential targets for interfering agents (endocrine disruptors) as well as for pharmacological interventions. Inactivating mutations have been identified as the underlying cause of heritable diseases (monocarboxylate transporter 8-associated Allan-Herndon-Dudley syndrome) and might also define a class of subclinical TH insensitivity. As a basic tool to solve questions regarding THTT substrate specificity, activation or inactivation by compounds and functional changes from mutations, uptake assays with radiolabeled tracers are standard. Due to the need for radioactive isotopes, this technique is limited to screening of labelled substrates and disadvantageous regarding handling, setup, and regulatory issues. To overcome these hurdles, we developed an uptake assay protocol using nonradioactive ligands. In brief, uptake of nonradioactive iodine-containing substrate molecules was monitored via Sandell-Kolthoff reaction. The novel assay was designed to the common microtiter plate layout. As a prove-of-principle, we measured TH uptake by monocarboxylate transporter 8-transfected MDCK1 cells. Titrations with bromosulphthalein as an example for inhibitor screening setups and a side-by-side comparison with the radioactive method prove this assay to be reliable, sensitive, and convenient. Furthermore, the method was applicable on primary murine astrocytes, which enables high-throughput screening studies on in vitro model systems with physiological transporter regulation. Due to its design, it is applicable for high-throughput screening of modulatory compounds, but it is also a safe, inexpensive and an easily accessible method for functional testing of THTT in basic science.