EMAIL THIS PAGE TO A FRIEND
Developmental biology
AVE protein expression and visceral endoderm cell behavior during anterior-posterior axis formation in mouse embryos: Asymmetry in OTX2 and DKK1 expression.
PMID 25910836
Abstract
The initial landmark of anterior-posterior (A-P) axis formation in mouse embryos is the distal visceral endoderm, DVE, which expresses a series of anterior genes at embryonic day 5.5 (E5.5). Subsequently, DVE cells move to the future anterior region, generating anterior visceral endoderm (AVE). Questions remain regarding how the DVE is formed and how the direction of the movement is determined. This study compares the detailed expression patterns of OTX2, HHEX, CER1, LEFTY1 and DKK1 by immunohistology and live imaging at E4.5-E6.5. At E6.5, the AVE is subdivided into four domains: most anterior (OTX2, HHEX, CER1-low/DKK1-high), anterior (OTX2, HHEX, CER1-high/DKK1-low), main (OTX2, HHEX, CER1, LEFTY1-high) and antero-lateral and posterior (OTX2, HHEX-low). The study demonstrates how this pattern is established. AVE protein expression in the DVE occurs de novo at E5.25-E5.5. Neither HHEX, LEFTY1 nor CER1 expression is asymmetric. In contrast, OTX2 expression is tilted on the future posterior side with the DKK1 expression at its proximal domain; the DVE cells move in the opposite direction of the tilt.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B8436 Anti-Brachyury antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB1406811
Anti-CER1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA019917
Anti-CER1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1411646
Anti-CER1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
D3195
Anti-DKK1 (N-terminal) antibody produced in rabbit, ~1.5 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB2500314
Anti-DKK1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
AV48015
Anti-DKK1 antibody produced in rabbit, IgG fraction of antiserum
HPA018995
Anti-DKK1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV39509
Anti-FOXA2 (AB2) antibody produced in rabbit, IgG fraction of antiserum
SAB2100836
Anti-FOXA2 antibody produced in rabbit, affinity isolated antibody
SAB2105304
Anti-FOXA2 antibody produced in rabbit, affinity isolated antibody
SAB2105303
Anti-FOXA2, (C-terminal) antibody produced in rabbit, affinity isolated antibody
AV38100
Anti-GATA4 antibody produced in rabbit, affinity isolated antibody
SAB4501128
Anti-GATA4 antibody produced in rabbit, affinity isolated antibody
SAB4501129
Anti-GATA4 antibody produced in rabbit, affinity isolated antibody
SAB4501130
Anti-GATA4 antibody produced in rabbit, affinity isolated antibody
AV32374
Anti-LHX1 (AB1) antibody produced in rabbit, affinity isolated antibody
AV100851
Anti-LHX1 (AB2) antibody produced in rabbit, IgG fraction of antiserum
SAB1412493
ANTI-LHX1 antibody produced in mouse, clone 1C11, purified immunoglobulin, buffered aqueous solution
SAB1412506 ANTI-LHX1 antibody produced in mouse, clone 3G3, purified immunoglobulin, buffered aqueous solution
SAB1412507 ANTI-LHX1 antibody produced in mouse, clone 2G5, purified immunoglobulin, buffered aqueous solution
SAB1412508 ANTI-LHX1 antibody produced in mouse, clone 1F1, purified immunoglobulin, buffered aqueous solution
SAB1412509 ANTI-LHX1 antibody produced in mouse, clone 4A5, purified immunoglobulin, buffered aqueous solution
SAB1412510
ANTI-LHX1 antibody produced in mouse, clone 2G10, purified immunoglobulin, buffered aqueous solution
SAB1412511 ANTI-LHX1 antibody produced in mouse, clone 4G5, purified immunoglobulin, buffered aqueous solution
SAB1412537
ANTI-LHX1 antibody produced in mouse, clone 2A7, purified immunoglobulin, buffered aqueous solution
SAB1412538
ANTI-LHX1 antibody produced in mouse, clone 2A8, purified immunoglobulin, buffered aqueous solution
SAB1412565 ANTI-LHX1 antibody produced in mouse, clone 2D7, purified immunoglobulin, buffered aqueous solution
SAB2102378
Anti-TBX1 antibody produced in rabbit, affinity isolated antibody
SAB4502925
Anti-TBX1, C-Terminal antibody produced in rabbit, affinity isolated antibody