EMAIL THIS PAGE TO A FRIEND
Journal of bioscience and bioengineering
Optimal fusion of antibody binding domains resulted in higher affinity and wider specificity.
PMID 25910963
Abstract
Antibody is a very important protein in biotechnological and biomedical fields because of its high affinity and specificity to various antigens. Due to the rise of human antibody therapeutics, its cost-effective purification is an urgent issue for bio-industry. In this study, we made novel fusion proteins PAxPG with a flexible (DDAKK)n linker between the two Ig binding domains derived from Staphylococcus protein A and Streptococcus protein G. The fusion proteins bound human and mouse IgGs and their fragments with up to 58-times higher affinity and wider specificity than the parental binding domains. Interestingly, the optimal linker for human Fab fragment was nxa0=xa04, which was close to the modeled distance between the termini of domains bound to heavy chain, implying increased avidity as a possible mechanism. For binding to Fc, the longest n=6 linker gave the highest affinity, implying longer interchain distance between the two binding sites. The novel fusion protein with optimized interdomain linker length will be a useful tool for the purification and detection of various IgGs including mouse IgG1 that binds only weakly to natural protein A.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P50404
1,3-Propanediol, 98%
C3H8O2
AV48225
Anti-VIM (AB1) antibody produced in rabbit, affinity isolated antibody
AV48226
Anti-VIM (AB2) antibody produced in rabbit, affinity isolated antibody
SAB1406585
Anti-VIM antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA001762
Anti-VIM antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4300676
Anti-Vimentin antibody produced in rabbit, affinity isolated antibody
SAB4503081
Anti-Vimentin antibody produced in rabbit, affinity isolated antibody
SAB4503083
Anti-Vimentin, C-Terminal antibody produced in rabbit, affinity isolated antibody
129585
Maleimide, 99%
C4H3NO2
WH0007431M1
Monoclonal Anti-VIM antibody produced in mouse, clone 3E9, purified immunoglobulin, buffered aqueous solution
SAB4700524 Monoclonal Anti-VIM antibody produced in mouse, clone VI-RE/1, purified immunoglobulin, buffered aqueous solution
SAB4100612
Monoclonal Anti-VIM antibody produced in mouse, clone VIM-A604, culture supernatant
AMAB90516
Monoclonal Anti-VIM antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0157, purified immunoglobulin, buffered aqueous glycerol solution
PLA0199
Rabbit anti-Vimentin Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.