EMAIL THIS PAGE TO A FRIEND
Molecular and cellular biochemistry
Cryptotanshinone, a Stat3 inhibitor, suppresses colorectal cancer proliferation and growth in vitro.
PMID 25912550
Abstract
Cryptotanshinone (CPT) is a natural compound extracted from herbal medicine that has been previously shown to possess antitumor properties in various types of human cancer cells. In the present study, we examined the potential role of CPT in the treatment of colorectal cancer. Using SW480, HCT116, and LOVO colorectal cancer cell lines, the effects of CPT on cell viability, apoptosis, and tumorigenicity were evaluated. The results showed that CPT significantly inhibited the growth and viability of SW480, HCT116, and LOVO cell lines by inducing apoptosis and prevented anchorage dependent growth on agar. In addition, CPT inhibited the activation of Signal transducer and activator of transcription 3 (Stat3) pathways in colorectal cancer cells. Stat3 is a transcription factor that mediates the expression of various genes associated with many cellular processes, such as inflammation and cell growth, and has been shown to promote several cancer types, including colorectal cancer. These findings indicate that CPT may be a potential candidate for the treatment and prevention of colorectal cancer in part by inhibiting the activation of Stat3.