EMAIL THIS PAGE TO A FRIEND
Archives of oral biology
Genomic identification and quantification of microbial species adhering to toothbrush bristles after disinfection: A cross-over study.
PMID 25912552
Abstract
The aim of this clinical investigation was to identify and quantify the microbial species adhering to toothbrush bristles after controlled brushing and storage in different antimicrobial agents. Sixteen healthy participants were enrolled in this study and randomly submitted to 4 interventions in a cross-over design: brushing and toothbrush storage in (I) Periogard/(II) Periobio (Chlorhexidine gluconate 0.12%), (III) Cepacol (cetylpyridinium chloride 0.05%) and (IV) distilled water (positive control). Thirty-eight bacterial species including putative pathogens and 5 Candida spp. were assessed by Checkerboard DNA-DNA hybridization. The results of the study have shown a striking reduction of the total microbial counts, including bacteria and Candida spp., on the toothbrush bristles after storage in cetylpyridinium chloride 0.05% (p < 0.0001). Chlorhexidine gluconate 0.12% showed no differences on the total bacterial count when compared to distilled water (p > 0.05). Cetylpyridinium chloride solution also presented the lowest genome counts and frequency of detection for individual target species; distilled water showed the highest individual genome counts (p < 0.05). Potential pathogenic species were recorded in moderate to high levels for chlorhexidine gluconate and distilled water. Cetylpyridinium chloride 0.05% was the most effective storage solution in the reduction of total and individual microbial counts, including pathogenic species.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C0732
Cetylpyridinium chloride, meets USP testing specifications
C21H38ClN · H2O