EMAIL THIS PAGE TO A FRIEND
Antonie van Leeuwenhoek
Novosphingobium tardum sp. nov., isolated from sediment of a freshwater lake.
PMID 25912732
Abstract
A Gram-negative, non-motive, aerobic and non-spore-forming strain 16-28-2(T) isolated from freshwater sediment of Taihu Lake was characterized by using a polyphasic approach. The optimum growth conditions were found to be as follows: 28 °C, pH 6.5 and 0-0.5 % NaCl in YG liquid medium. The major fatty acids were identified to be summed feature 3 (consisting of C16:1 ω7c and/or C16:1 ω6c), summed feature 8 (consisting of C18:1 ω7c and/or C18:1 ω6c), C14:0 2-OH, C17:1 ω6c, C16:0 and C18:1 ω7c 11-methyl (>5 %). Strain 16-28-2(T) was found to contain diphosphatidylglycerol, phosphatidylglycerol, phosphatidylethanolamine, phosphatidylcholine and sphingoglycolipid as the major polar lipids; and ubiquinone 10 (Q-10) as the major respiratory quinone. DNA G+C content of strain 16-28-2(T) was 63.5 mol % (Tm). A phylogenetic study of 16S rRNA gene indicated that strain 16-28-2(T) is a member of the genus Novosphingobium, with the highest 16S rRNA gene sequence similarity of 96.3 % with Novosphingobium lentum MT1(T) and below 96 % with the other Novosphingobium species. On the basis of the phylogenetic, phenotypic analyses and biochemical characterization, we suggest that strain 16-28-2(T) is a novel species in the genus Novosphingobium, for which the name Novosphingobium tardum sp. nov. is proposed. The type strain of N. tardum is 16-28-2(T) (=CGMCC 1.12989(T) =NBRC 110956(T)).