EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
Identification of the absorbed components and metabolites of Zhi-Zi-Da-Huang decoction in rat plasma by ultra-high performance liquid chromatography coupled with quadrupole-time-of-flight mass spectrometry.
PMID 25912849
Abstract
Zhi-Zi-Da-Huang decoction (ZZDHD), consisting of Gardenia jasminoides Ellis, Rheum palmatum L., Citrus aurantium L. and Sojae Semen Praeparatum, is a widely used traditional Chinese medicine preparation for the treatment of acute or chronic hepatic diseases. In the present study, a sensitive and selective ultra-high performance liquid chromatography coupled with quadrupole-time-of-flight mass spectrometry (UHPLC-Q-TOF-MS) method was developed to separate and identify the absorbed components and metabolites in rat plasma after oral administration of ZZDHD. The plasma samples were pretreated by protein precipitation and separated on a Shim-pack XR-ODS C18 column (75 mm × 3.0 mm, 2.2 μm) using a gradient elution program. Mass spectrometric detection was performed on an Agilent 6520 Q-TOF mass spectrometer equipped with electrospray ionization (ESI) source in positive and negative ion modes. By comparing the retention time, high resolution mass data of blank plasma and dosed plasma, a total of 43 constituents, including 21 prototype compounds and 22 metabolites were identified or tentatively characterized. Results indicated that glucuronidation and sulfation were the main metabolic pathways of iridoid glycosides and anthraquinones, glucuronidation was the main metabolic pathways of flavanone-related compounds. It is concluded the developed UHPLC-Q-TOF-MS method with high sensitivity and resolution is suitable for identifying and characterizing the absorbed components and metabolites of ZZDHD, and the results will provide essential data for further studying the relationship between the chemical components and pharmacological activity of ZZDHD.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SMB00153
Asperulosidic acid, ≥90% (LC/MS-ELSD)
C18H24O12