EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
NDR1 modulates the UV-induced DNA-damage checkpoint andxa0nucleotide excision repair.
PMID 25912875
Abstract
Nucleotide excision repair (NER) is the sole mechanism of UV-induced DNA lesion repair in mammals. A single round of NER requires multiple components including seven core NER factors, xeroderma pigmentosum A-G (XPA-XPG), and many auxiliary effector proteins including ATR serine/threonine kinase. The XPA protein helps to verify DNA damage and thus plays a rate-limiting role in NER. Hence, the regulation of XPA is important for the entire NER kinetic. We found that NDR1, a novel XPA-interacting protein, modulates NER by modulating the UV-induced DNA-damage checkpoint. In quiescent cells, NDR1 localized mainly in the cytoplasm. After UV irradiation, NDR1 accumulated in the nucleus. The siRNA knockdown of NDR1 delayed the repair of UV-induced cyclobutane pyrimidine dimers in both normal cells and cancer cells. It did not, however, alter the expression levels or the chromatin association levels of the core NER factors following UV irradiation. Instead, the NDR1-depleted cells displayed reduced activity of ATR for some set of its substrates including CHK1 and p53, suggesting that NDR1 modulates NER indirectly via the ATR pathway.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GW22272A Anti-GC4 (ab1) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
GW22272B Anti-GC4 (ab2) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
AV47974
Anti-NDRG1 (AB1) antibody produced in rabbit, IgG fraction of antiserum
AV47975
Anti-NDRG1 (AB2) antibody produced in rabbit, IgG fraction of antiserum
N8539
Anti-NDRG1 (C-terminal) antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB2500677
Anti-NDRG1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2701820
Anti-ndrg1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA006881
Anti-NDRG1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2501750
Anti-STK38 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2501883
Anti-STK38 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
AV48727
Anti-STK38 antibody produced in rabbit, affinity isolated antibody
SAB1410182
Anti-STK38 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
X1254 Anti-XPA (C-terminal) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
X1504 Anti-XPA (N-terminal) antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB1406599
Anti-XPA antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1411400
Anti-XPA antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4503090
Anti-XPA, C-Terminal antibody produced in rabbit, affinity isolated antibody
WH0010397M3
Monoclonal Anti-NDRG1 antibody produced in mouse, clone 2D7, purified immunoglobulin, buffered aqueous solution
WH0011329M1
Monoclonal Anti-STK38 antibody produced in mouse, clone 2G8-1F3, purified immunoglobulin, buffered aqueous solution
SAB1408832
Monoclonal Anti-STK38 antibody produced in mouse, clone 3A5, purified immunoglobulin, buffered aqueous solution
SAB1408833
Monoclonal Anti-STK38 antibody produced in mouse, clone 6F1, purified immunoglobulin, buffered aqueous solution
SAB1408834
Monoclonal Anti-STK38 antibody produced in mouse, clone 2F3, purified immunoglobulin, buffered aqueous solution
SAB1408835
Monoclonal Anti-STK38 antibody produced in mouse, clone 6G11, purified immunoglobulin, buffered aqueous solution