EMAIL THIS PAGE TO A FRIEND
Respiratory medicine
Airway epithelial cytokine responses in childhood wheeze are independent of atopic status.
PMID 25912933
Abstract
Airway epithelial cells (AEC) are key contributors to immune function in the lungs but little is known about their role and function in children. Having previously established that nasal AEC mediator release correlates with that of bronchial AEC, we assessed AEC responses in children with and without a history of wheeze. Nasal AEC cultures were established from children (0.6-14.9 years) undergoing elective surgical procedures under general anaesthetic categorised as atopic asthmatic (nxa0=xa012), virus-induced wheeze (nxa0=xa08) or children without wheeze (nxa0=xa032). Mediator release by AEC monolayers at passage 2 was determined by cytometric bead array assay or ELISA. Unstimulated AEC from children with a history of wheeze produced significantly less IL-8, IL-6, MCP-1 and G-CSF than AEC from healthy controls. There were no group differences in AEC release of VEGF, RANTES, MMP-9 or TIMP-1. After stimulation with the pro-inflammatory cytokines IL-1β and TNFα, AEC from children with current wheeze produced significantly less IL-8, IL-6 and MCP-1 than children without wheeze. Release of G-CSF, VEGF, MMP-9 and TIMP-1 did not differ between the wheeze and control group. There were no differences in mediator release between subjects with atopic asthma and those with virus-induced wheeze or between atopic and non-atopic controls. On multivariate analysis, wheeze was the only significant predictor of AEC mediator release. Intrinsic differences in AEC from children with a history of wheeze may reflect a defect in cytokine production inxa0vivo or an altered state of differentiation inxa0vitro, independent of atopic status.