EMAIL THIS PAGE TO A FRIEND
Proteomics
Unravelling the proteome of degenerative human mitral valves.
PMID 25914152
Abstract
Degenerative mitral valve disease (DMVD), which includes the syndromes of mitral valve prolapse (MVP) and flail leaflet, is a common valvular condition which can be complicated by mitral regurgitation and adverse cardiovascular outcomes. Although several genetic and other studies of MVP in dog models have provided some information regarding the underlying disease mechanisms, the proteins and molecular events mediating human MVP pathogenesis have not been unraveled. In this study, we report the first large-scale proteome profiling of mitral valve tissue resected from patients with MVP. A total of 1134 proteins were identified, some of which were validated using SWATH-MS and western blotting. GO annotation of these proteins confirmed the validity of this proteome database in various cardiovascular processes. Among the list of proteins, we found several structural and extracellular matrix proteins, such as asporin, biglycan, decorin, lumican, mimecan, prolargin, versican, and vinculin, that have putative roles in the pathophysiology of MVP. These proteins could also be involved in the cardiac remodeling associated with mitral regurgitation. All MS data have been deposited in the ProteomeXchange with identifier PXD000774 (http://proteomecentral.proteomexchange.org/dataset/PXD000774).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA007583
Anti-COL3A1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1408562
Anti-COL3A1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1301234
ANTI-COMP (CENTER) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1411896
ANTI-COMP antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA000848
Anti-FBLN5 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA000868
Anti-FBLN5 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4500114
Anti-FBLN5 antibody produced in rabbit, affinity isolated antibody
SAB1409292
Anti-LUM antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA001522
Anti-LUM antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1401232
Anti-LUM antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2107069
Anti-LUM antibody produced in rabbit, affinity isolated antibody
SAB3701093 Anti-Mouse IgG (H+L) antibody produced in sheep, affinity isolated antibody, buffered aqueous solution
SAB1406201
Anti-OGN antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA013132
Anti-OGN antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1408612
Anti-OGN antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1400223
Anti-PRELP antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
SAB2103061
Anti-PRELP antibody produced in rabbit, affinity isolated antibody
SAB1408906
Anti-VCAN antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA004726 Anti-VCAN antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB5300101
Monoclonal Anti-FBLN5 antibody produced in mouse, clone 3F10A5, ascites fluid
SAB5300102
Monoclonal Anti-FBLN5 antibody produced in mouse, clone 3F8D5, ascites fluid
SAB5300103
Monoclonal Anti-FBLN5 antibody produced in mouse, clone 1G6A4, ascites fluid
SAB1402861
Monoclonal Anti-OGN, (C-terminal) antibody produced in mouse, clone 2G4, purified immunoglobulin, buffered aqueous solution
SAB1402879
Monoclonal Anti-PRELP antibody produced in mouse, clone 3H1, ascites fluid