EMAIL THIS PAGE TO A FRIEND
Arthritis & rheumatology (Hoboken, N.J.)
A Role for Gut Microbiota and the Metabolite-Sensing Receptor GPR43 in a Murine Model of Gout.
PMID 25914377
Abstract
Host-microbial interactions are central in health and disease. Monosodium urate monohydrate (MSU) crystals cause gout by activating the NLRP3 inflammasome, leading to interleukin-1β (IL-1β) production and neutrophil recruitment. This study was undertaken to investigate the relevance of gut microbiota, acetate, and the metabolite-sensing receptor GPR43 in regulating inflammation in a murine model of gout. Gout was induced by the injection of MSU crystals into the knee joints of mice. Macrophages from the various animals were stimulated to determine inflammasome activation and production of reactive oxygen species (ROS). Injection of MSU crystals caused joint inflammation, as seen by neutrophil influx, hypernociception, and production of IL-1β and CXCL1. These parameters were greatly decreased in germ-free mice, mice treated with antibiotics, and GPR-43-deficient mice. Recolonization or administration of acetate to germ-free mice restored inflammation in response to injection of MSU crystals. In vitro, macrophages produced ROS and assembled the inflammasome when stimulated with MSU. Macrophages from germ-free animals produced little ROS, and there was little inflammasome assembly. Similar results were observed in macrophages from GPR-43-deficient mice. Treatment of germ-free mice with acetate restored in vitro responsiveness of macrophages to MSU crystals. In the absence of microbiota, there is decreased production of short-chain fatty acids that are necessary for adequate inflammasome assembly and IL-1β production in a manner that is at least partially dependent on GPR43. These results clearly show that the commensal microbiota shapes the host's ability to respond to an inflammasome-dependent acute inflammatory stimulus outside the gut.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
FLAAS
Adenosine 5′-triphosphate (ATP) disodium salt hydrate, vial of ~1 mg ATP
C10H14N5Na2O13P3 · xH2O
A1852
Adenosine 5′-triphosphate (ATP) disodium salt hydrate, vial of 30 mg
C10H14N5Na2O13P3 · xH2O
A7699
Adenosine 5′-triphosphate disodium salt hydrate, BioXtra, ≥99% (HPLC), from microbial
C10H14N5Na2O13P3 · xH2O
A26209
Adenosine 5′-triphosphate disodium salt hydrate, 99%
C10H14N5Na2O13P3 · xH2O
A2383
Adenosine 5′-triphosphate disodium salt hydrate, Grade I, ≥99%, from microbial
C10H14N5Na2O13P3 · xH2O
A3377
Adenosine 5′-triphosphate disodium salt hydrate, Grade II, ≥98.5% (HPLC), crystalline, from microbial
C10H14N5Na2O13P3 · xH2O
A6419
Adenosine 5′-triphosphate disodium salt hydrate, microbial, BioReagent, suitable for cell culture, ≥99% (HPLC)
C10H14N5Na2O13P3 · xH2O
S2889
Sodium acetate, anhydrous, for molecular biology, ≥99%
C2H3NaO2
S3272
Sodium acetate, for electrophoresis, ≥99%, crystalline
C2H3NaO2
S5636
Sodium acetate, powder, BioReagent, for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
C2H3NaO2
S1429
Sodium acetate, meets USP testing specifications, anhydrous
C2H3NaO2
229873
Sodium acetate, 99.995% trace metals basis
C2H3NaO2
S7545
Sodium acetate, BioXtra, ≥99.0%
C2H3NaO2
71183
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
C2H3NaO2
S8388
Sodium acetate solution, 0.3 M
C2H3NaO2
71196
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
C2H3NaO2
U2625
Uric acid, ≥99%, crystalline
C5H4N4O3
U0881
Uric acid, BioXtra, ≥99% (HPLC)
C5H4N4O3