EMAIL THIS PAGE TO A FRIEND
International immunopharmacology
Gastroprotective effect of andrographolide sodium bisulfite against indomethacin-induced gastric ulceration in rats.
PMID 25916678
Abstract
Andrographolide sodium bisulfite (ASB), a water-soluble sulfonate of andrographolide has been shown to possess anti-inflammatory, antipyretic and analgesic activities. However, there is no report on the gastroprotective effect of ASB against indomethacin-induced gastric ulcer. Here we investigated the possible anti-ulcerogenic potential of ASB and the underlying mechanism against indomethacin-induced gastric ulcer in rats. The ulcer area, histopathological assessment, contents of gastric mucosal glutathione (GSH), catalase (CAT), superoxide dismutase (SOD), malonaldehyde (MDA) and prostaglandin E2 (PGE2) were examined. In addition, cyclooxygenase-1 (COX-1), cyclooxygenase-2 (COX-2) mRNA expression and immunohistochemical evaluation of HSP70, Bcl-2 and Bax proteins were also investigated. Results indicated that ASB pre-treatment significantly reduced the ulcer areas induced by indomethacin compared with the vehicle group. The gastric levels of GSH, CAT and SOD were markedly increased by ASB while the level of MDA was decreased. In addition, ASB pretreatment significantly promoted the gastric PGE2 levels and up-regulated the COX-1 and COX-2 mRNA expression in comparison with the vehicle group. Immunohistochemistry analysis showed obvious up-regulation of HSP70 and Bcl-2 protein expression while suppression of Bax protein in the gastric tissue of ASB-pretreated group. Taken together, these findings indicated that the gastroprotective effect of ASB might be associated with the improvement of antioxidative status, activation of COX-mediated PGE2 synthesis, down-regulation of Bax proteins and up-regulation of Bcl-2 and HSP70 proteins. ASB might have the potential for further development as a promising alternative for antiulcer treatment.