EMAIL THIS PAGE TO A FRIEND
Cancer letters
Smac mimetic and oleanolic acid synergize to induce cell death in human hepatocellular carcinoma cells.
PMID 25917078
Abstract
Chemotherapy resistance of hepatocellular carcinoma (HCC) is still a major unsolved problem highlighting the need to develop novel therapeutic strategies. Here, we identify a novel synergistic induction of cell death by the combination of the Smac mimetic BV6, which antagonizes Inhibitor of apoptosis (IAP) proteins, and the triterpenoid oleanolic acid (OA) in human HCC cells. Importantly, BV6 and OA also cooperate to suppress long-term clonogenic survival as well as tumor growth in a preclinical in vivo model of HCC underscoring the clinical relevance of our findings. In contrast, BV6/OA cotreatment does not exert cytotoxic effects against normal primary hepatocytes, pointing to some tumor selectivity. Mechanistic studies show that BV6/OA cotreatment leads to DNA fragmentation and caspase-3 cleavage, while supply of the pan-caspase inhibitor N-benzyloxycarbonyl-Val-Ala-Asp-fluoromethylketone (zVAD.fmk) revealed a cell type-dependent requirement of caspases for BV6/OA-induced cell death. The receptor interacting protein (RIP)1 kinase Inhibitor Necrostatin-1 (Nec-1) or genetic knockdown of RIP1 fails to rescue BV6/OA-mediated cell death, indicating that BV6/OA cotreatment does not primarily engage necroptotic cell death. Notably, the addition of several reactive oxygen species (ROS) scavengers significantly decreases BV6/OA-triggered cell death, indicating that ROS production contributes to BV6/OA-induced cell death. In conclusion, cotreatment of Smac mimetic and OA represents a novel approach for the induction of cell death in HCC and implicates further studies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA001302
Anti-CASP8 antibody produced in rabbit, Ab3, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA005688
Anti-CASP8 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA001473
Anti-CASP9 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4503332
Anti-CASP9 antibody produced in rabbit, affinity isolated antibody
SAB4503333
Anti-CASP9 antibody produced in rabbit, affinity isolated antibody
SAB4503334
Anti-CASP9 antibody produced in rabbit, affinity isolated antibody
SAB4503335
Anti-CASP9 antibody produced in rabbit, affinity isolated antibody
GW21154 Anti-Caspase 8 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
C7729 Anti-Caspase 9 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4300683
Anti-Caspase 9 antibody produced in rabbit, affinity isolated antibody
SAB3500404
Anti-Caspase-8 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3500405
Anti-Caspase-9 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA021335
Anti-ENDOG antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
HPA021830
Anti-ENDOG antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
SAB2103518
Anti-ENDOG antibody produced in rabbit, affinity isolated antibody
SAB1405757
Anti-ENDOG, (C-terminal) antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
E5654
Anti-Endonuclease G antibody produced in rabbit, 1 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB5300353 Monoclonal Anti-CASP8 antibody produced in mouse, clone 1H11, ascites fluid
WH0000842M1
Monoclonal Anti-CASP9 antibody produced in mouse, clone 3B8-4G2, purified immunoglobulin, buffered aqueous solution
SAB1403634
Monoclonal Anti-CASP9 antibody produced in mouse, clone 4F12-G6, purified immunoglobulin, buffered aqueous solution
C4106 Monoclonal Anti-Caspase 8 antibody produced in mouse, ~2 mg/mL, clone CAS8, purified immunoglobulin, buffered aqueous solution
C4356
Monoclonal Anti-Caspase 9 antibody produced in mouse, ~2 mg/mL, clone CAS9, purified immunoglobulin, buffered aqueous solution