EMAIL THIS PAGE TO A FRIEND
Acta biomaterialia
Hydrosoluble, UV-crosslinkable and injectable chitosan for patterned cell-laden microgel and rapid transdermal curing hydrogel in vivo.
PMID 25917845
Abstract
Natural and biodegradable chitosan with unique amino groups has found widespread applications in tissue engineering and drug delivery. However, its applications have been limited by the poor solubility of native chitosan in neutral pH solution, which subsequently fails to achieve cell-laden hydrogel at physiological pH. To address this, we incorporated UV crosslinking ability in chitosan, allowing fabrication of patterned cell-laden and rapid transdermal curing hydrogel in vivo. The hydrosoluble, UV crosslinkable and injectable N-methacryloyl chitosan (N-MAC) was synthesized via single-step chemoselective N-acylation reaction, which simultaneously endowed chitosan with well solubility in neutral pH solution, UV crosslinkable ability and injectability. The solubility of N-MAC in neutral pH solution increased 2.21-fold with substitution degree increasing from 10.9% to 28.4%. The N-MAC allowed fabrication of cell-laden microgels with on-demand patterns via photolithography, and the cell viability in N-MAC hydrogel maintained 96.3 ± 1.3% N-MAC allowed rapid transdermal curing hydrogel in vivo within 60s through minimally invasive clinical surgery. Histological analysis revealed that low-dose UV irradiation hardly induced skin injury and acute inflammatory response disappeared after 7 days. N-MAC would allow rapid, robust and cost-effective fabrication of patterned cell-laden polysaccharide microgels with unique amino groups serving as building blocks for tissue engineering and rapid transdermal curing hydrogel in vivo for localized and sustained protein delivery.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
276685
Methacrylic anhydride, contains 2,000 ppm topanol A as inhibitor, 94%
C8H10O3