EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Human NOTCH2 Is Resistant to Ligand-independent Activation by Metalloprotease Adam17.
PMID 25918160
Abstract
Cell surface receptors of the NOTCH family of proteins are activated by ligand induced intramembrane proteolysis. Unfolding of the extracellular negative regulatory region (NRR), enabling successive proteolysis by the enzymes Adam10 and γ-secretase, is rate-limiting in NOTCH activation. Mutations in the NOTCH1 NRR are associated with ligand-independent activation and frequently found in human T-cell malignancies. In mammals four NOTCH receptors and five Delta/Jagged ligands exist, but mutations in the NRR are only rarely reported for receptors other than NOTCH1. Using biochemical and functional assays, we compared the molecular mechanisms of ligand-independent signaling in NOTCH1 and the highly related NOTCH2 receptor. Both murine Notch1 and Notch2 require the metalloprotease protease Adam17, but not Adam10 during ligand-independent activation. Interestingly, the human NOTCH2 receptor is resistant to ligand-independent activation compared with its human homologs or murine orthologs. Taken together, our data reveal subtle but functionally important differences for the NRR among NOTCH paralogs and homologs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1405933
Anti-HIF1A antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA001275
Anti-HIF1A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2101039
Anti-HIF1A antibody produced in rabbit, affinity isolated antibody
SAB2104366
Anti-HIF1A antibody produced in rabbit, affinity isolated antibody
SAB2702132
Anti-HIF1A antibody produced in rabbit
L1293
Anti-Lamin A (C-terminal) antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4501766
Anti-Lamin A (Cleaved-Asp230) antibody produced in rabbit, affinity isolated antibody
SAB4501767
Anti-Lamin A (Cleaved-Asp230) antibody produced in rabbit, affinity isolated antibody
SAB4501764
Anti-Lamin A antibody produced in rabbit, affinity isolated antibody
SAB1406087
Anti-LMNA antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA006660
Anti-LMNA antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4502019
Anti-NOTCH1 (Cleaved-Val1754) antibody produced in rabbit, affinity isolated antibody
N4788 Anti-NOTCH1 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB2101618
Anti-NOTCH1 antibody produced in rabbit, affinity isolated antibody
SAB4502021
Anti-NOTCH2 (Cleaved-Ala1734) antibody produced in rabbit, affinity isolated antibody
SAB4502020
Anti-NOTCH2 (Cleaved-Asp1733) antibody produced in rabbit, affinity isolated antibody
SAB4502022
Anti-NOTCH2 (Cleaved-Val1697) antibody produced in rabbit, affinity isolated antibody
N4913 Anti-NOTCH2 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB2101619
Anti-NOTCH2 antibody produced in rabbit, affinity isolated antibody
HPA046392
Anti-NOTCH2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
PLA0304
Goat anti-HIF1-alpha Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
SAB5200017
Monoclonal Anti-HIF-1 α antibody produced in mouse, clone ESEE122, 1 mg/mL, purified immunoglobulin
H6536 Monoclonal Anti-HIF-1α antibody produced in mouse, ~1 mg/mL, clone H1α67, purified immunoglobulin, buffered aqueous solution
H6411 Monoclonal Anti-HIF-1α antibody produced in mouse, ~1 mg/mL, clone OZ12, purified immunoglobulin, buffered aqueous solution
SAB1403916
Monoclonal Anti-HIF1A, (C-terminal) antibody produced in mouse, clone 1D4, purified immunoglobulin, buffered aqueous solution
SAB4200236
Monoclonal Anti-Lamin A/C antibody produced in mouse, clone 4C11, purified from hybridoma cell culture
SAB4700742 Monoclonal Anti-Notch 1 antibody produced in mouse, clone mN1A, purified immunoglobulin, buffered aqueous solution
SAB4200024 Monoclonal Anti-Notch 1 antibody produced in rat, ~1.0 mg/mL, clone ICN 5B5, purified immunoglobulin, buffered aqueous solution
N6786
Monoclonal Anti-Notch1 antibody produced in mouse, clone mN1A, purified immunoglobulin, buffered aqueous solution
SAB3300078
Monoclonal Anti-Notch1 antibody produced in mouse, clone 3E12, purified immunoglobulin, buffered aqueous glycerol solution