EMAIL THIS PAGE TO A FRIEND
Journal of immunology research
Mesenchymal stem cells from patients with rheumatoid arthritis display impaired function in inhibiting Th17 cells.
PMID 25918734
Abstract
Mesenchymal stem cells (MSCs) possess multipotent and immunomodulatory properties and are suggested to be involved in the pathogenesis of immune-related diseases. This study explored the function of bone marrow MSCs from rheumatoid arthritis (RA) patients, focusing on immunomodulatory effects. RA MSCs showed decreased proliferative activity and aberrant migration capacity. No significant differences were observed in cytokine profiles between RA and control MSCs. The effects of RA MSCs on proliferation of peripheral blood mononuclear cells (PBMCs) and distribution of specific CD4(+) T cell subtypes (Th17, Treg, and Tfh cells) were investigated. RA MSCs appeared to be indistinguishable from controls in suppressing PBMC proliferation, decreasing the proportion of Tfh cells, and inducing the polarization of Treg cells. However, the capacity to inhibit Th17 cell polarization was impaired in RA MSCs, which was related to the low expression of CCL2 in RA MSCs after coculture with CD4(+) T cells. These findings indicated that RA MSCs display defects in several important biological activities, especially the capacity to inhibit Th17 cell polarization. These functionally impaired MSCs may contribute to the development of RA disease.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
21888
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
C29H19NO11
87444
5-Carboxy-fluorescein diacetate N-succinimidyl ester, for fluorescence, ≥95.0% (HPLC)
C29H19NO11
A9400
7-Aminoactinomycin D, ~97% (HPLC), powder
C62H87N13O16
SAB1406413
Anti-CCL2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA019163
Anti-CCL2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2103318
Anti-CCL2 antibody produced in rabbit, affinity isolated antibody
SAB1411231
Anti-CCL2 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2501381
Anti-Cd28 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2501674
Anti-CD28 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1405583
Anti-CD28 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
C7831 Anti-CD28 antibody, Mouse monoclonal, clone CD28.2, purified from hybridoma cell culture
SAB1412683
ANTI-IFNA1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1410692
Anti-IFNA1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
AV54329
Anti-IL4 antibody produced in rabbit, affinity isolated antibody
SAB2103750
Anti-IL4 antibody produced in rabbit, affinity isolated antibody
I7776 Anti-Interleukin-4 antibody produced in goat, affinity isolated antibody, lyophilized powder
I7526 Anti-Interleukin-4 antibody produced in goat, affinity isolated antibody, lyophilized powder
I8784 Anti-Interleukin-4 antibody produced in goat, IgG fraction of antiserum, lyophilized powder
B7651
Brefeldin A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
C16H24O4
B6542
Brefeldin A, ≥99% (HPLC and TLC), BioXtra, for molecular biology
C16H24O4
B5936
Brefeldin A, from Penicillium brefeldianum, Ready Made Solution, 10 mg/mL in DMSO, 0.2 μm filtered
C16H24O4
03609
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
C10H16N2O8
03620
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
C10H16N2O8
03690
Ethylenediaminetetraacetic acid solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
C10H16N2O8
SAB1402908
Monoclonal Anti-CCL2 antibody produced in mouse, clone 1F10, purified immunoglobulin, buffered aqueous solution
SAB1409632
Monoclonal Anti-CCL2 antibody produced in mouse, clone 2E2, purified immunoglobulin, buffered aqueous solution
SAB4700135 Monoclonal Anti-CD28 antibody produced in mouse, low endotoxin, clone CD28.2, purified immunoglobulin, buffered aqueous solution
SAB4700739 Monoclonal Anti-CD28 antibody produced in mouse, clone JJ319, purified immunoglobulin, buffered aqueous solution
SAB4700751
Monoclonal Anti-CD28 antibody produced in mouse, clone CD28.2, purified immunoglobulin, buffered aqueous solution
WH0003439M7
Monoclonal Anti-IFNA1 antibody produced in mouse, clone 3C6, purified immunoglobulin, buffered aqueous solution