EMAIL THIS PAGE TO A FRIEND
Blood
STAT1 is phosphorylated and downregulated by the oncogenic tyrosine kinase NPM-ALK in ALK-positive anaplastic large-cell lymphoma.
PMID 25921060
Abstract
The tumorigenicity of most cases of ALK-positive anaplastic large-cell lymphoma (ALK+ ALCL) is driven by the oncogenic fusion protein NPM-ALK in a STAT3-dependent manner. Because it has been shown that STAT3 can be inhibited by STAT1 in some experimental models, we hypothesized that the STAT1 signaling pathway is defective in ALK+ ALCL, thereby leaving the STAT3 signaling unchecked. Compared with normal T cells, ALK+ ALCL tumors consistently expressed a low level of STAT1. Inhibition of the ubiquitin-proteasome pathway appreciably increased STAT1 expression in ALK+ ALCL cells. Furthermore, we found evidence that NPM-ALK binds to and phosphorylates STAT1, thereby promoting its proteasomal degradation in a STAT3-dependent manner. If restored, STAT1 is functionally intact in ALK+ ALCL cells, because it effectively upregulated interferon-γ, induced apoptosis/cell-cycle arrest, potentiated the inhibitory effects of doxorubicin, and suppressed tumor growth in vivo. STAT1 interfered with the STAT3 signaling by decreasing STAT3 transcriptional activity/DNA binding and its homodimerization. The importance of the STAT1/STAT3 functional interaction was further highlighted by the observation that short interfering RNA knockdown of STAT1 significantly decreased apoptosis induced by STAT3 inhibition. Thus, STAT1 is a tumor suppressor in ALK+ ALCL. Phosphorylation and downregulation of STAT1 by NPM-ALK represent other mechanisms by which this oncogenic tyrosine kinase promotes tumorigenesis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1105169 Anti-STAT1 (671-685) antibody produced in rabbit, IgG fraction of antiserum
SAB4300326
Anti-STAT1 (Ab-701) antibody produced in rabbit, affinity isolated antibody
SAB4300441
Anti-STAT1 (Ab-727) antibody produced in rabbit, affinity isolated antibody
GW22758 Anti-STAT1 (ab1) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
AV33373
Anti-STAT1 (AB1) antibody produced in rabbit, affinity isolated antibody
GW22780 Anti-STAT1 (ab2) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
AV33374
Anti-STAT1 (AB2) antibody produced in rabbit, affinity isolated antibody
AV38933
Anti-STAT1 (AB3) antibody produced in rabbit, affinity isolated antibody
AV38934
Anti-STAT1 (AB4) antibody produced in rabbit, affinity isolated antibody
SAB1406486
Anti-STAT1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA000931
Anti-STAT1 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV36979
Anti-STAT1 antibody produced in rabbit, IgG fraction of antiserum
S5683 Anti-STAT1 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous glycerol solution
HPA000982
Anti-STAT1 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4502868
Anti-STAT1, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4502869
Anti-STAT1, C-Terminal antibody produced in rabbit, affinity isolated antibody
WH0006772M1
Monoclonal Anti-STAT1 antibody produced in mouse, clone 1A8, purified immunoglobulin, buffered aqueous solution