EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
The synthesis of new amphiphilic p-tert-butylthiacalix[4]arenes containing peptide fragments and their interaction with DNA.
PMID 25921225
Abstract
New water-soluble p-tert-butylthiacalix[4]arenes containing peptide and quaternary ammonium fragments in cone and 1,3-alternate conformations were synthesized and characterized. The interaction of the macrocycles with DNA was studied by UV-spectroscopy, DLS and TEM. It was shown that the interaction of the self-associates based on p-tert-butylthiacalix[4]arenes tetrasubstituted at the lower rim with glycine and quaternary ammonium fragments in cone and 1,3-alternate conformations with DNA led to the formation of particles of about 99-192 nm in size.