EMAIL THIS PAGE TO A FRIEND
Journal of ethnopharmacology
Astragaloside IV and cycloastragenol are equally effective in inhibition of endoplasmic reticulum stress-associated TXNIP/NLRP3 inflammasome activation in the endothelium.
PMID 25922268
Abstract
Astragaloside IV and cycloastragenol are present together in Astragalus membranaceus Moench (Fabaceae) and this study aims to simultaneously investigate their regulation of endothelial homeostasis in the setting of endoplasmic reticulum stress (ER stress). We stimulated endothelial cells with palmitate (PA 100μM) to evoked ROS-associated ER stress and observed the effects of astragaloside IV and cycloastragenol on thioredoxin-interacting protein (TXNIP) expression, NLRP3 inflammasome activation and mitochondrion-dependent apoptosis. Astragaloside IV and cycloastragenol inhibited ROS generation and attenuated ER stress inducer IRE1α phosphorylation, indicating the inhibition of ROS-associated ER stress. In response to ER stress, TXNIP expression increased, accompanied with NLRP3 induction and increased IL-1β and IL-6 production, but these alternations were reversed by treatment with astragaloside IV and cycloastragenol, demonstrating the inhibitory effects of astragaloside IV and cycloastragenol on TXNIP/NLRP3 inflammasome activation. Inflammasome activation led to mitochondrial cell death in endothelial cells, whereas astragaloside IV and cycloastragenol restored the loss of the mitochondrial membrane potential with inhibition of caspase-3 activity, and thereby protected cells from ER stress-induced apoptosis. Astragaloside IV and cycloastragenol enhanced AMPK phosphorylation and AMPK inhibitor compound C diminished their beneficial effects, indicative of the potential role of AMPK in their regulation. Astragaloside IV and cycloastragenol suppressed ROS-associated ER stress and then inhibited TXNIP/NLRP3 inflammasome activation with regulation of AMPK activity, and thereby ameliorated endothelial dysfunction by inhibiting inflammation and reducing cell apoptosis. Simultaneous investigations further showed that astragaloside IV and cycloastragenol were equally effective in regulation of endothelial homeostasis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
164968
5-Amino-4-imidazolecarboxamide hydrochloride, 98%
C4H6N4O · HCl
SAB1412027
ANTI-ERN1 antibody produced in mouse, clone 1F12, purified immunoglobulin, buffered aqueous solution
SAB1412028
ANTI-ERN1 antibody produced in mouse, clone 5F3, purified immunoglobulin, buffered aqueous solution
SAB1412029
ANTI-ERN1 antibody produced in mouse, clone 6G11, purified immunoglobulin, buffered aqueous solution
SAB1412341
ANTI-ERN1 antibody produced in mouse, clone 4E1, purified immunoglobulin, buffered aqueous solution
SAB1412645
ANTI-ERN1 antibody produced in mouse, clone 4D12, purified immunoglobulin, buffered aqueous solution
HPA027730 Anti-ERN1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
PRS3655
Anti-IRE1P (ab1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS3657
Anti-IRE1P (ab2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS3659
Anti-IRE1P (ab3) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
T6528
Anti-Mouse IgG (Fab specific)–TRITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
T7782 Anti-Mouse IgG (Fab specific)–TRITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3701031 Anti-Mouse IgG (Fc specific), F(ab′)2 fragment, highly cross adsorbed-Biotin antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3701028 Anti-Mouse IgG (Fc specific), F(ab′)2 fragment, highly cross adsorbed-Fluorescein antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3701011 Anti-Mouse IgG (Fc specific), F(ab′)2 fragment, highly cross adsorbed-Rhodamine antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3701033 Anti-Mouse IgG (Fc specific), F(ab′)2 fragment, highly cross adsorbed-Texas Red™ antibody produced in goat, affinity isolated antibody, lyophilized powder
B7401 Anti-Mouse IgG (Fc specific)–Biotin antibody produced in goat, affinity isolated antibody, buffered aqueous solution
B9904 Anti-Mouse IgG (Fc specific)–Biotin antibody produced in goat, affinity isolated antibody, buffered aqueous solution
B7022 Anti-Mouse IgG (γ-chain specific)−Biotin antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB4600013
Anti-Mouse IgG (H+L), CF 350 antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600019
Anti-Mouse IgG (H+L), CF 405M antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600023
Anti-Mouse IgG (H+L), CF 405S antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600042
Anti-Mouse IgG (H+L), CF 488A antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600066
Anti-Mouse IgG (H+L), CF 555 antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600082
Anti-Mouse IgG (H+L), CF 568 antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600105
Anti-Mouse IgG (H+L), CF 594 antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600138
Anti-Mouse IgG (H+L), CF 633 antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600161
Anti-Mouse IgG (H+L), CF 640R antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600182
Anti-Mouse IgG (H+L), CF 647 antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600191
Anti-Mouse IgG (H+L), CF 660C antibody produced in goat, ~2 mg/mL, affinity isolated antibody