EMAIL THIS PAGE TO A FRIEND
Oncotarget
Diet-induced hypercholesterolemia promotes androgen-independent prostate cancer metastasis via IQGAP1 and caveolin-1.
PMID 25924234
Abstract
Obesity and metabolic syndrome are associated with several cancers, however, the molecular mechanisms remain to be fully elucidated. Recent studies suggest that hypercholesterolemia increases intratumoral androgen signaling in prostate cancer, but it is unclear whether androgen-independent mechanisms also exist. Since hypercholesterolemia is associated with advanced, castrate-resistant prostate cancer, in this study, we aimed to determine whether and how hypercholesterolemia affects prostate cancer progression in the absence of androgen signaling. We demonstrate that diet-induced hypercholesterolemia promotes orthotopic xenograft PC-3 cell metastasis, concomitant with elevated expression of caveolin-1 and IQGAP1 in xenograft tumor tissues. In vitro cholesterol treatment of PC-3 cells stimulated migration and increased IQGAP1 and caveolin-1 protein level and localization to a detergent-resistant fraction. Down-regulation of caveolin-1 or IQGAP1 in PC-3 cells reduced migration and invasion in vitro, and hypercholesterolemia-induced metastasis in vivo. Double knock-down of caveolin-1 and IQGAP1 showed no additive effect, suggesting that caveolin-1 and IQGAP1 act via the same pathway. Taken together, our data show that hypercholesterolemia promotes prostate cancer metastasis independent of the androgen pathway, in part by increasing IQGAP1 and caveolin-1. These results have broader implications for managing metastasis of cancers in general as IQGAP1 and hypercholesterolemia are implicated in the progression of several cancers.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2900061
Anti-CAV1 (AB1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2900062
Anti-CAV1 (AB2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
AV09019
Anti-CAV1 antibody produced in rabbit, purified antibody, Protein A purified
SAB2900063
Anti-CAV1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2100344
Anti-CAV1 antibody produced in rabbit, affinity isolated antibody
C4490
Anti-Caveolin-1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
C3237
Anti-Caveolin-1 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB4503427
Anti-Caveolin-1, C-Terminal antibody produced in rabbit, affinity isolated antibody
HPA005928
Anti-EMCN antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2700787
Anti-EMCN antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701919 Anti-flot1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA001393
Anti-FLOT1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410072
Anti-FLOT1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2500404
Anti-Flotillin 1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
F1180
Anti-Flotillin 1 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB2501549
Anti-IQGAP1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA014055
Anti-IQGAP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4200329
Anti-IQGAP1 antibody produced in rabbit, ~1.5 mg/mL, affinity isolated antibody
SAB4502163
Anti-Mucin-14, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4200216
Monoclonal Anti-Caveolin-1 antibody produced in mouse, clone CAV1, tissue culture supernatant
SAB1403056
Monoclonal Anti-FLOT1 antibody produced in mouse, clone 4A1, ascites fluid
WH0008826M1
Monoclonal Anti-IQGAP1 antibody produced in mouse, clone 2C5, purified immunoglobulin, buffered aqueous solution
PLA0225
Rabbit anti-IQGAP1 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
T1500
Testosterone, ≥98%
C19H28O2
T-037
Testosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material
C19H28O2