EMAIL THIS PAGE TO A FRIEND
American journal of physiology. Renal physiology
Effect of dietary protein restriction on renal ammonia metabolism.
PMID 25925252
Abstract
Dietary protein restriction has multiple benefits in kidney disease. Because protein intake is a major determinant of endogenous acid production, it is important that net acid excretion change in parallel during protein restriction. Ammonia is the primary component of net acid excretion, and inappropriate ammonia excretion can lead to negative nitrogen balance. Accordingly, we examined ammonia excretion in response to protein restriction and then we determined the molecular mechanism of the changes observed. Wild-type C57Bl/6 mice fed a 20% protein diet and then changed to 6% protein developed an 85% reduction in ammonia excretion within 2 days, which persisted during a 10-day study. The expression of multiple proteins involved in renal ammonia metabolism was altered, including the ammonia-generating enzymes phosphate-dependent glutaminase (PDG) and phosphoenolpyruvate carboxykinase (PEPCK) and the ammonia-metabolizing enzyme glutamine synthetase. Rhbg, an ammonia transporter, increased in expression in the inner stripe of outer medullary collecting duct intercalated cell (OMCDis-IC). However, collecting duct-specific Rhbg deletion did not alter the response to protein restriction. Rhcg deletion did not alter ammonia excretion in response to dietary protein restriction. These results indicate 1) dietary protein restriction decreases renal ammonia excretion through coordinated regulation of multiple components of ammonia metabolism; 2) increased Rhbg expression in the OMCDis-IC may indicate a biological role in addition to ammonia transport; and 3) Rhcg expression is not necessary to decrease ammonia excretion during dietary protein restriction.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1307110
ANTI-GLS2 (C-TERM E513) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
AV43562
Anti-GLS2 antibody produced in rabbit, IgG fraction of antiserum
SAB3500720
Anti-GLS2 antibody produced in rabbit, affinity isolated antibody
SAB2701668
Anti-glul antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701726
Anti-glul antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA007316
Anti-GLUL antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA007571
Anti-GLUL antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1408580
Anti-GLUL antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
G2781
Anti-Glutamine Synthetase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
HPA018107
Anti-SLC12A1 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA014967
Anti-SLC12A1 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV41388
Anti-SLC12A1 antibody produced in rabbit, affinity isolated antibody
SAB5200103
Anti-SLC12A1 antibody produced in rabbit, 1 mg/mL, affinity isolated antibody
HPA036493
Anti-SLC9A3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA036669
Anti-SLC9A3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB5200102
Anti-SLC9A3 antibody produced in rabbit, 1 mg/mL, affinity isolated antibody
WH0002752M1
Monoclonal Anti-GLUL antibody produced in mouse, clone 2B12, purified immunoglobulin, buffered aqueous solution
SAB1403872
Monoclonal Anti-GLUL, (C-terminal) antibody produced in mouse, clone 3B6, purified immunoglobulin, buffered aqueous solution
SAB1305525
MONOCLONAL ANTI-LGA (CENTER) antibody produced in mouse, clone 585CT26.3.1, crude ascites, buffered aqueous solution
SAB1402357
Monoclonal Anti-SLC12A1 antibody produced in mouse, clone 4H4, purified immunoglobulin, buffered aqueous solution