EMAIL THIS PAGE TO A FRIEND
Biology of reproduction
Regulatory Role of Gonadotropins and Local Factors Produced by Ovarian Follicles on In Vitro Resistin Expression and Action on Porcine Follicular Steroidogenesis.
PMID 25926437
Abstract
Resistin, a hormone secreted by adipocytes, is thought to be important in reproduction. Our previous study demonstrated resistin expression in porcine ovarian follicles and its direct effect on steroidogenesis. The aim of the current study was to evaluate the effect of gonadotropins and the local ovarian factors, such as insulin-like growth factor type 1 (IGF1) and steroids (progesterone, testosterone, and 17 beta-estradiol), on the expression and secretion of resistin, as well as its steroidogenic action. Porcine ovarian follicles were exposed to follicle-stimulating hormone (FSH) and luteinizing hormone (LH) at 50-150 ng/ml, IGF1 (10-100 ng/ml), and steroids at 10(-8) to 10(-6) M for 24 h. Then, mRNA, protein expression, and medium concentration of resistin were determined using real-time PCR, Western blot analysis, and ELISA, respectively. In the subsequent experiments, ovarian follicles were exposed to resistin and/or FSH, LH, IGF1, and steroids, and ovarian steroidogenesis was analyzed. Additionally, we examined the direct effect of resistin on the protein expression of receptors for gonadotropins and investigated local factors. The results showed that gonadotropins and steroids have stimulatory effects but that IGF1 has an inhibitory effect on resistin expression and secretion. Resistin decreased gonadotropins and local hormone-induced steroid secretion and inhibited 3beta-hydroxysteroid dehydrogenase, 17beta-hydroxysteroid dehydrogenase, and cytochrome P450 aromatase protein expression. Additionally, we demonstrated that resistin increased the expression of receptors for progesterone and testosterone. These findings all show that the expression and function of resistin are regulated by gonadotropins and local factors produced by ovarian follicles.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B9673
1-Bromo-3-chloropropane, for isolation of RNA
C3H6BrCl
B62404
1-Bromo-3-chloropropane, 99%
C3H6BrCl
SAB4300348
Anti-PGR (Ab-190) antibody produced in rabbit, affinity isolated antibody
HPA004751
Anti-PGR antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV100687
Anti-PGR antibody produced in rabbit, affinity isolated antibody
SAB2101781
Anti-PGR antibody produced in rabbit, affinity isolated antibody
HPA008428
Anti-PGR antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2105280
Anti-PGR antibody produced in rabbit, affinity isolated antibody
HPA017176
Anti-PGR antibody produced in rabbit, Ab3, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
GW22679 Anti-Progesterone Receptor antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB4502184
Anti-Progesterone Receptor antibody produced in rabbit, affinity isolated antibody
SAB4502185
Anti-Progesterone Receptor antibody produced in rabbit, affinity isolated antibody
SAB4502186
Anti-Progesterone Receptor antibody produced in rabbit, affinity isolated antibody
HPA053794
Anti-TMEM37 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
F2293 Follicle Stimulating Hormone from porcine pituitary
SRP4121 IGF-I from rat, recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC)
SRP3069 IGF-I human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), cell culture tested
I3769 Insulin-like Growth Factor-I human, IGF-I, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
L5269 Luteinizing Hormone from sheep pituitary, lyophilized powder
WH0005241M1
Monoclonal Anti-PGR antibody produced in mouse, clone 3E11, purified immunoglobulin, buffered aqueous solution
SAB1400206
Monoclonal Anti-PGR antibody produced in mouse, clone 5D10, purified immunoglobulin, buffered aqueous solution
SAB1400207
Monoclonal Anti-PGR antibody produced in mouse, clone 4E9, purified immunoglobulin, buffered aqueous solution
SAB1404196
Monoclonal Anti-PGR, (N-terminal) antibody produced in mouse, clone 2C2, purified immunoglobulin, buffered aqueous solution