EMAIL THIS PAGE TO A FRIEND
Journal of virology
New Noncoding Lytic Transcripts Derived from the Epstein-Barr Virus Latency Origin of Replication, oriP, Are Hyperedited, Bind the Paraspeckle Protein, NONO/p54nrb, and Support Viral Lytic Transcription.
PMID 25926645
Abstract
We have previously shown that the Epstein-Barr virus (EBV) likely encodes hundreds of viral long noncoding RNAs (vlncRNAs) that are expressed during reactivation. Here we show that the EBV latency origin of replication (oriP) is transcribed bi-directionally during reactivation and that both leftward (oriPtLs) and rightward (oriPtRs) transcripts are largely localized in the nucleus. While the oriPtLs are most likely noncoding, at least some of the oriPtRs contain the BCRF1/vIL10 open reading frame. Nonetheless, oriPtR transcripts with long 5' untranslated regions may partially serve noncoding functions. Both oriPtL and oriPtR transcripts are expressed with late kinetics, and their expression is inhibited by phosphonoacetic acid. RNA sequencing (RNA-seq) analysis showed that oriPtLs and oriPtRs exhibited extensive "hyperediting" at their Family of Repeat (FR) regions. RNA secondary structure prediction revealed that the FR region of both oriPtLs and oriPtRs may form large evolutionarily conserved and thermodynamically stable hairpins. The double-stranded RNA-binding protein and RNA-editing enzyme ADAR was found to bind to oriPtLs, likely facilitating editing of the FR hairpin. Further, the multifunctional paraspeckle protein, NONO, was found to bind to oriPt transcripts, suggesting that oriPts interact with the paraspeckle-based innate antiviral immune pathway. Knockdown and ectopic expression of oriPtLs showed that it contributes to global viral lytic gene expression and viral DNA replication. Together, these results show that these new vlncRNAs interact with cellular innate immune pathways and that they help facilitate progression of the viral lytic cascade. Recent studies have revealed that the complexity of lytic herpesviral transcriptomes is significantly greater than previously appreciated with hundreds of viral long noncoding RNAs (vlncRNAs) being recently discovered. Work on cellular lncRNAs over the past several years has just begun to give us an initial appreciation for the array of functions they play in complex formation and regulatory processes in the cell. The newly identified herpesvirus lncRNAs are similarly likely to play a variety of different functions, although these functions are likely tailored to specific needs of the viral infection cycles. Here we describe novel transcripts derived from the EBV latency origin of replication. We show that they are hyperedited, that they interact with a relatively newly appreciated antiviral pathway, and that they play a role in facilitating viral lytic gene expression. These investigations are a starting point to unraveling the complex arena of vlncRNA function in herpesvirus lytic replication.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1405425
Anti-ADAR antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1401004
Anti-ADAR antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4200542
Anti-ADAR1 (C-terminal) antibody produced in rabbit, IgG fraction of antiserum
SAB4200541
Anti-ADAR1 antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
SAB4500077
Anti-ADAR1, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB2502094
Anti-CHRM2 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1405619
Anti-CHRM2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA029795
Anti-CHRM2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4500245
Anti-CHRM2 antibody produced in rabbit, affinity isolated antibody
I2136 Anti-Human IgG (Fc specific) antibody produced in goat, affinity isolated antibody, buffered aqueous solution
I0759 Anti-Human IgM (μ-chain specific) antibody produced in goat, affinity isolated antibody, lyophilized powder
M9693 Anti-Muscarinic Acetylcholine Receptor M2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
AMAB90535
Monoclonal Anti-ADAR antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0176, purified immunoglobulin, buffered aqueous glycerol solution
SAB4200482
Monoclonal Anti-ADAR1 antibody produced in mouse, ~1.0 mg/mL, clone ADAR61, purified immunoglobulin
WH0001129M1
Monoclonal Anti-CHRM2 antibody produced in mouse, clone 4B5, purified immunoglobulin, buffered aqueous solution
S6508
Sodium orthovanadate, ≥90% (titration)
Na3O4V
450243
Sodium orthovanadate, 99.98% trace metals basis
Na3O4V