PloS one
The shh signaling pathway is upregulated in multiple cell types in cortical ischemia and influences the outcome of stroke in an animal model.
PMID 25927436
Abstract
Recently the sonic hedgehog (shh) signaling pathway has been shown to play an important role in regulating repair and regenerative responses after brain injury, including ischemia. However, the precise cellular components that express and upregulate the shh gene and the cellular components that respond to shh signaling remain to be identified. In this study, using a distal MCA occlusion model, our data show that the shh signal is upregulated both at the cortical area near the injury site and in the adjacent striatum. Multiple cell types upregulate shh signaling in ischemic brain, including neurons, reactive astrocytes and nestin-expressing cells. The shh signaling pathway genes are also expressed in the neural stem cells (NSCs) niche in the subventricular zone (SVZ). Conditional deletion of the shh gene in nestin-expressing cells both at the SVZ niche and at the ischemic site lead to significantly more severe behavioral deficits in these shh iKO mice after cortical stroke, measured using an automated open field locomotion apparatus (Student's t-test, p<0.05). In contrast, animals given post-stroke treatment with the shh signaling agonist (SAG) demonstrated less deficits in behavioral function, compared to vehicle-treated mice. At 7 days after stroke, SAG-treated mice showed higher values in multiple horizontal movement parameters compared to vehicle treated mice (Student's t-test, p<0.05) whereas there were no differences in pre-stroke measurements, (Student's t-test, p>0.05). In summary, our data demonstrate that shh signaling plays critical and ongoing roles in response to ischemic injury and modulation of shh signaling in vivo alters the functional outcome after cortical ischemic injury.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4500629
Anti-Doublecortin, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB2501172 Anti-ptc1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1411779
ANTI-PTCH antibody produced in mouse, clone 1D9, purified immunoglobulin, buffered aqueous solution
SAB1411866
ANTI-PTCH antibody produced in mouse, clone 8A10, purified immunoglobulin, buffered aqueous solution
SAB1411867
ANTI-PTCH antibody produced in mouse, clone 1B7, purified immunoglobulin, buffered aqueous solution
SAB1411868
ANTI-PTCH antibody produced in mouse, clone 3B7, purified immunoglobulin, buffered aqueous solution
SAB1411869
ANTI-PTCH antibody produced in mouse, clone 3C2, purified immunoglobulin, buffered aqueous solution
SAB1411871
ANTI-PTCH antibody produced in mouse, clone 3B3, purified immunoglobulin, buffered aqueous solution
SAB1411872
ANTI-PTCH antibody produced in mouse, clone 4F11, purified immunoglobulin, buffered aqueous solution
SAB2501613 Anti-PTCH1 (C-terminal) antibody produced in goat, affinity isolated antibody, buffered aqueous solution
AV44249
Anti-PTCH1 antibody produced in rabbit, affinity isolated antibody
AV44235
Anti-SHH antibody produced in rabbit, affinity isolated antibody
SAB1412475
ANTI-SMO antibody produced in mouse, clone 2D10, purified immunoglobulin, buffered aqueous solution
SAB1412476
ANTI-SMO antibody produced in mouse, clone 4E5, purified immunoglobulin, buffered aqueous solution
SAB1412477
ANTI-SMO antibody produced in mouse, clone 4E9, purified immunoglobulin, buffered aqueous solution
SAB2500961
Anti-SMO, C-Terminal antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1101510
Anti-SMOX (151-165) antibody produced in rabbit, IgG fraction of antiserum
SAB1101511
Anti-SMOX (401-415) antibody produced in rabbit, IgG fraction of antiserum
HPA047117
Anti-SMOX antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
S8321
Anti-Sonic Hedgehog antibody, Mouse monoclonal, clone SH154, purified from hybridoma cell culture
WH0005727M2
Monoclonal Anti-PTCH antibody produced in mouse, clone 1G2, purified immunoglobulin, buffered aqueous solution
SAB1404271
Monoclonal Anti-PTCH antibody produced in mouse, clone 8E1, purified immunoglobulin, buffered aqueous solution
WH0006469M3
Monoclonal Anti-SHH antibody produced in mouse, clone 1B11, purified immunoglobulin, buffered aqueous solution
WH0006608M1 Monoclonal Anti-SMO antibody produced in mouse, clone 1D6, purified immunoglobulin, buffered aqueous solution
SAB1404382
Monoclonal Anti-SMO, (C-terminal) antibody produced in mouse, clone 1D9, purified immunoglobulin, buffered aqueous solution
S4944 Monoclonal Anti-Sonic Hedgehog (N-terminal) antibody produced in rat, clone 171018, purified immunoglobulin, lyophilized powder