EMAIL THIS PAGE TO A FRIEND
Cell cycle (Georgetown, Tex.)
IQGAP1 is associated with nuclear envelope reformation and completion of abscission.
PMID 25928398
Abstract
The final stage of mitosis is cytokinesis, which results in 2 independent daughter cells. Cytokinesis has 2 phases: membrane ingression followed by membrane abscission. IQGAP1 is a scaffold protein that interacts with proteins implicated in mitosis, including F-actin, myosin and CaM. IQGAP1 in yeast recruits actin and myosin II filaments to the contractile ring for membrane ingression. In contrast, we show that mammalian IQGAP1 is not required for ingression, but coordinates nuclear pore complex (NPC) reassembly and completion of abscission. Depletion of IQGAP1 disrupts Nup98 and mAb414 nuclear envelope localization and delays abscission timing. IQGAP1 phosphorylation increases 15-fold upon mitotic entry at S86, S330 and T1434, with the latter site being targeted by CDK2/Cyclin A and CDK1/Cyclin A/B in vitro. Expressing the phospho-deficient mutant IQGAP1-S330A impairs NPC reassembly in cells undergoing abscission. Thus, mammalian IQGAP1 functions later in mitosis than its yeast counterpart to regulate nuclear pore assembly in a S330 phosphorylation-dependent manner during the abscission phase of cytokinesis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2100037
Anti-ACTB antibody produced in rabbit, affinity isolated antibody
GW23014 Anti-Actin, Cytoplasmic 1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB2501631
Anti-AURKC (N-terminal) antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1406495
Anti-AURKC antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2103462
Anti-AURKC antibody produced in rabbit, affinity isolated antibody
HPA034859
Anti-AURKC antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
A5102 Anti-Aurora B antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1300093
Anti-Aurora-C (C-term) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1300369
Anti-Aurora-C (center) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1300366
Anti-Aurora-C (N-term) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB3500350
Anti-β-Actin antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB2501549
Anti-IQGAP1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA014055
Anti-IQGAP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4200329
Anti-IQGAP1 antibody produced in rabbit, ~1.5 mg/mL, affinity isolated antibody
SAB1409385
Anti-NUP98 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV53631
Anti-NUP98 antibody produced in rabbit, affinity isolated antibody
SAB2101665
Anti-NUP98 antibody produced in rabbit, affinity isolated antibody
SAB1410851
Anti-NUP98 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
WH0000060M1
Monoclonal Anti-ACTB antibody produced in mouse, clone 3G4-F9, purified immunoglobulin, buffered aqueous solution
SAB1403520
Monoclonal Anti-ACTB antibody produced in mouse, clone 1E7, purified immunoglobulin, buffered aqueous solution
WH0009212M3
Monoclonal Anti-AURKB antibody produced in mouse, clone 6H7, purified immunoglobulin, buffered aqueous solution
SAB5300517
Monoclonal Anti-AURKB antibody produced in mouse, clone 13E8D3, ascites fluid
SAB1404685
Monoclonal Anti-AURKB, (N-terminal) antibody produced in mouse, clone 6G8, purified immunoglobulin, buffered aqueous solution
WH0008826M1
Monoclonal Anti-IQGAP1 antibody produced in mouse, clone 2C5, purified immunoglobulin, buffered aqueous solution
WH0004928M1 Monoclonal Anti-NUP98 antibody produced in mouse, clone 4G2, purified immunoglobulin, buffered aqueous solution
N1038
Monoclonal Anti-Nup98 antibody produced in rat, ~2 mg/mL, clone 2H10, purified immunoglobulin, buffered aqueous solution
PLA0225
Rabbit anti-IQGAP1 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.