EMAIL THIS PAGE TO A FRIEND
Vox sanguinis
Protocol guided bleeding management improves cardiac surgery patient outcomes.
PMID 25930098
Abstract
Excessive bleeding is a risk associated with cardiac surgery. Treatment invariably requires transfusion of blood products; however, the transfusion itself may contribute to postoperative sequelae. Our objective was to analyse a quality initiative designed to provide an evidenced-based approach to bleeding management. A retrospective analysis compared blood product transfusion and patient outcomes 15 months before and after implementation of a bleeding management protocol. The protocol incorporated point-of-care coagulation testing (POCCT) with ROTEM and Multiplate to diagnose the cause of bleeding and monitor treatment. Use of the protocol led to decreases in the incidence of transfusion of PRBCs (47·3% vs. 32·4%; P < 0·0001), FFP (26·9% vs. 7·3%; P < 0·0001) and platelets (36·1% vs. 13·5%; P < 0·0001). During the intra-operative period, the percentage of patients receiving cryoprecipitate increased (2·7% vs. 5·1%; P = 0·002), as did the number of units transfused (248 vs. 692; P < 0·0001). The proportion of patients who received tranexamic acid increased (13·7% to 68·2%; P < 0·0001). There were reductions in re-exploration for bleeding (5·6% vs. 3·4; P = 0·01), superficial chest wound (3·3% vs. 1·4%; P = 0·002), leg wound infection (4·6% vs. 2·0%; P < 0·0001) and a 12% reduction in mean length of stay from operation to discharge (95%: 9-16%, P < 0·0001). Acquisition cost of blood products decreased by $1 029 118 in the 15-month period with the protocol. The implementation of a bleeding management protocol supported by POCCT in a cardiac surgery programme was associated with significant reductions in the transfusion of allogeneic blood products, improved outcomes and reduced cost.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
857653
trans-4-(Aminomethyl)cyclohexanecarboxylic acid, 97%
C8H15NO2