EMAIL THIS PAGE TO A FRIEND
Placenta
Maternal diabetes induces changes in the umbilical cord gene expression.
PMID 25935091
Abstract
Since maternal diabetes may affect fetal development and the umbilical cord provides an extension of the fetal vasculature, we decided to investigate cords' biological responses to maternal diabetic milieu. Using microarray analysis, we determined the gene expression profiles in the umbilical cords of six neonates born to type 1 diabetic mothers and in six control cords. Umbilical cord tissue was collected immediately after elective cesarean section. Expression data were confirmed by real-time polymerase chain reaction (11 genes). Additionally, the same umbilical cords were analyzed histologically. Two hundred eighty six genes were differentially expressed in the umbilical cords from diabetic pregnancies compared to the controls (fold change ±1.5 and Pxa0