EMAIL THIS PAGE TO A FRIEND
Acta diabetologica
Fenofibrate prevents the disruption of the outer blood retinal barrier through downregulation of NF-κB activity.
PMID 25936740
Abstract
There is clinical evidence that fenofibrate, a PPARα agonist, arrests the progression of diabetic macular edema (DME). However, the underlying mechanisms of this beneficial effect remain to be elucidated. We previously reported that fenofibric acid (FA), the active metabolite of fenofibrate, prevents the disorganization of tight junction proteins and the hyperpermeability provoked by the diabetic milieu in the retinal pigment epithelium (RPE). The aim of the present study was to evaluate whether this effect is mediated by inhibiting the proinflammatory transcription factor NF-κB, as well as the expression of several proinflammatory cytokines involved in the pathogenesis of DME. Human RPE cells were cultured under standard conditions and under conditions leading to the disruption of the monolayer [IL-1β (10 ng/ml)]. The effect of FA, QNZ (a NF-κB inhibitor), WY14643 (a PPARα agonist), and MK-866 (a PPARα antagonist) in the disruption of the monolayer was determined by dextran permeability and immunohistochemistry analyses. The effect of FA on NF-κB activity was assessed by EMSA and by NF-κB/p65 nuclear translocation analyses. The expression of cytokines (IL-6, IL-8, MCP-1) was measured by RT-PCR. FA prevented RPE monolayer disruption, and the consequent hyperpermeability induced by IL-1β, through inhibition of NF-κB activity. This effect was due to PPARα activation and was associated with a significant downregulation of the expression of proinflammatory cytokines. Our findings suggest that the anti-inflammatory effects of FA through inhibition of NF-κB activity play a key role in the beneficial effect of fenofibrate for treating DME.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0579
EVP4593, ≥98% (HPLC)
C22H20N4O
C7081
WY-14643
C14H14ClN3O2S