EMAIL THIS PAGE TO A FRIEND
Osteoarthritis and cartilage
Heparin-binding epidermal growth factor-like growth factor (HB-EGF) is increased in osteoarthritis and regulates chondrocyte catabolic and anabolic activities.
PMID 25937027
Abstract
We determined if the epidermal growth factor receptor ligand HB-EGF is produced in cartilage and if it regulates chondrocyte anabolic or catabolic activity. HB-EGF expression was measured by quantitative PCR using RNA isolated from mouse knee joint tissues and from normal and osteoarthritis (OA) human chondrocytes. Immunohistochemistry was performed on normal and OA human cartilage and meniscus sections. Cultured chondrocytes were treated with fibronectin fragments (FN-f) as a catabolic stimulus and osteogenic protein 1 (OP-1) as an anabolic stimulus. Effects of HB-EGF on cell signaling were analyzed by immunoblotting of selected signaling proteins. MMP-13 was measured in conditioned media, proteoglycan synthesis was measured by sulfate incorporation, and matrix gene expression by quantitative PCR. HB-EGF expression was increased in 12-month old mice at 8 weeks after surgery to induce OA and increased amounts of HB-EGF were noted in human articular cartilage from OA knees. FN-f stimulated chondrocyte HB-EGF expression and HB-EGF stimulated chondrocyte MMP-13 production. However, HB-EGF was not required for FN-f stimulation of MMP-13 production. HB-EGF activated the ERK and p38 MAP kinases and stimulated phosphorylation of Smad1 at an inhibitory serine site which was associated with inhibition of OP-1 mediated proteoglycan synthesis and reduced aggrecan (ACAN) but not COL2A1 expression. HB-EGF is a new factor identified in OA cartilage that promotes chondrocyte catabolic activity while inhibiting anabolic activity suggesting it could contribute to the catabolic-anabolic imbalance seen in OA cartilage.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2104809
Anti-EGF antibody produced in rabbit, affinity isolated antibody, lyophilized powder
GW22768 Anti-MAPK1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
HPA030069
Anti-MAPK1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
M4065 Anti-Matrix Metalloproteinase-2 antibody produced in rabbit, ~1 mg/mL, IgG fraction of antiserum, buffered aqueous solution
SAB4501891
Anti-MMP-2, C-Terminal antibody produced in rabbit, affinity isolated antibody
GW21124 Anti-MMP13 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1406136
Anti-MMP13 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2104396
Anti-MMP13 antibody produced in rabbit, affinity isolated antibody
HPA001939
Anti-MMP2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV20016
Anti-MMP2 antibody produced in rabbit, IgG fraction of antiserum
RAB0149
Human EGF ELISA Kit, for serum, plasma, cell culture supernatant and urine
RAB0364
Human MMP-13 ELISA Kit, for serum, plasma, cell culture supernatant and urine
WH0001950M1
Monoclonal Anti-EGF antibody produced in mouse, clone 2F1, purified immunoglobulin, buffered aqueous solution
SAB5300488
Monoclonal Anti-EGF antibody produced in mouse, clone 9D7F11, ascites fluid
SAB5300489
Monoclonal Anti-EGF antibody produced in mouse, clone 4E11, ascites fluid
E2520 Monoclonal Anti-Epidermal Growth Factor antibody produced in mouse, clone EGF-10, purified immunoglobulin, buffered aqueous solution
SAB1305558 MONOCLONAL ANTI-ERK2 antibody produced in mouse, clone 814CT5.4.2, IgG fraction of antiserum, buffered aqueous solution
WH0005594M1
Monoclonal Anti-MAPK1 antibody produced in mouse, clone 1D1, purified immunoglobulin, buffered aqueous solution
WH0004322M7
Monoclonal Anti-MMP13 antibody produced in mouse, clone 3B11, purified immunoglobulin, buffered aqueous solution
RAB0150
Mouse EGF ELISA Kit, for serum, plasma and cell culture supernatant
PLA0236
Rabbit anti-ERK2 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
RAB0567
Rat Egf / Pro-epidermal Growth Factor ELISA Kit, for serum, plasma and cell culture supernatants