PloS one
Strategies to rescue the consequences of inducible arginase-1 deficiency in mice.
PMID 25938595
Abstract
Arginase-1 catalyzes the conversion of arginine to ornithine and urea, which is the final step of the urea cycle used to remove excess ammonia from the body. Arginase-1 deficiency leads to hyperargininemia in mice and man with severe lethal consequences in the former and progressive neurological impairment to varying degrees in the latter. In a tamoxifen-induced arginase-1 deficient mouse model, mice succumb to the enzyme deficiency within 2 weeks after inducing the knockout and retain <2 % enzyme in the liver. Standard clinical care regimens for arginase-1 deficiency (low-protein diet, the nitrogen-scavenging drug sodium phenylbutyrate, ornithine supplementation) either failed to extend lifespan (ornithine) or only minimally prolonged lifespan (maximum 8 days with low-protein diet and drug). A conditional, tamoxifen-inducible arginase-1 transgenic mouse strain expressing the enzyme from the Rosa26 locus modestly extended lifespan of neonatal mice, but not that of 4-week old mice, when crossed to the inducible arginase-1 knockout mouse strain. Delivery of an arginase-1/enhanced green fluorescent fusion construct by adeno-associated viral delivery (rh10 serotype with a strong cytomegalovirus-chicken β-actin hybrid promoter) rescued about 30% of male mice with lifespan prolongation to at least 6 months, extensive hepatic expression and restoration of significant enzyme activity in liver. In contrast, a vector of the AAV8 serotype driven by the thyroxine-binding globulin promoter led to weaker liver expression and did not rescue arginase-1 deficient mice to any great extent. Since the induced arginase-1 deficient mouse model displays a much more severe phenotype when compared to human arginase-1 deficiency, these studies reveal that it may be feasible with gene therapy strategies to correct the various manifestations of the disorder and they provide optimism for future clinical studies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV45673
Anti-ARG1 (AB2) antibody produced in rabbit, IgG fraction of antiserum
SAB2501375
Anti-ARG1 (C-terminal) antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1405480
Anti-ARG1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA003595
Anti-ARG1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA024006
Anti-ARG1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2100144
Anti-ARG1 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB4200510
Anti-ARG1 antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
SAB1407859
Anti-TINAGL1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2106148
Anti-TINAGL1 antibody produced in rabbit, affinity isolated antibody
SAB2106149
Anti-TINAGL1 antibody produced in rabbit, affinity isolated antibody
HPA048695
Anti-TINAGL1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
203025
Cesium chloride, ≥99.999% trace metals basis
ClCs
449733
Cesium chloride, anhydrous, beads, −10 mesh, 99.999% trace metals basis
ClCs
C3139
Cesium chloride, optical grade, ≥99.5% trace metals basis
ClCs
C3011
Cesium chloride, Grade I, ≥99.0%
ClCs
562599
Cesium chloride, 99.99% trace metals basis
ClCs
C3032
Cesium chloride, for molecular biology, ≥99% (silver nitrate titration)
ClCs
C4036
Cesium chloride, for molecular biology, ≥98%
ClCs
C3309
Cesium chloride, BioXtra, ≥99.5% (titration)
ClCs
C6914
Cesium chloride, Grade II, ≥98%
ClCs
20966
Cesium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
ClCs
SAB4200508
Monoclonal Anti-ARG1 antibody produced in mouse, ~1.0 mg/mL, clone ARG1-3, purified immunoglobulin
SAB4100251
Monoclonal Anti-ARG1 antibody produced in mouse, clone ARG1-A219, culture supernatant
SAB4200509
Monoclonal Anti-ARG1 antibody produced in mouse, ~1.0 mg/mL, clone ARG1-6, purified immunoglobulin
AMAB90545
Monoclonal Anti-ARG1 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0186, purified immunoglobulin, buffered aqueous glycerol solution