EMAIL THIS PAGE TO A FRIEND
Stem cells (Dayton, Ohio)
Sox2 Deacetylation by Sirt1 Is Involved in Mouse Somatic Reprogramming.
PMID 25940188
Abstract
Mouse somatic cells can be reprogrammed into induced pluripotent stem cells by defined factors known to regulate pluripotency, including Oct4, Sox2, Klf4, and c-Myc. Together with Oct4, Sox2 plays a major role as a master endogenous pluripotent genes trigger in reprogramming. It has been reported that Sirtuin 1 (Sirt1), a member of the Sirtuin family of NAD(+) -dependent protein deacetylases, is involved in embryonic stem cell antioxidation, differentiation, and individual development. However, as a deacetylation enzyme, whether Sirt1 influences reprogramming through its post-translational modification function remains unknown. In this study, we provide evidence that deacetylation of Sox2 by Sirt1 is required for reprogramming. We found that a low level of Sox2 acetylation could significantly increase reprogramming efficiency. Furthermore, we found that Sox2 can be deacetylated by Sirt1 in an Oct4-mediated manner. Compared with wild-type cells, Sirt1-null mouse embryonic fibroblasts exhibit decreased reprogramming efficiency, and overexpression of Sirt1 rescues this defect. In addition, Sirt1 functions in the regulation of reprogramming through deacetylating Sox2. Taken together, we have identified a new regulatory role of Sirt1 in reprogramming and provided a link between deacetylation events and somatic cell reprogramming. Stem Cells 2015;33:2135-2147.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S5313
Anti-Sir2 (AS-16) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
S5322
Anti-Sirt1 (C-terminal) antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB3500493
Anti-SIRT1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB2501994
Anti-SIRT1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
S5447
Anti-SIRT1 antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution
AV32386
Anti-SIRT1 antibody produced in rabbit, affinity isolated antibody
EHU021051 MISSION® esiRNA, esiRNA human MYC (esiRNA1)
EMU075291 MISSION® esiRNA, esiRNA targeting mouse Myc (esiRNA1)
S5196
Monoclonal Anti-Sirt1 antibody produced in mouse, ~2 mg/mL, clone SIR11, purified immunoglobulin, buffered aqueous solution
WH0023411M1
Monoclonal Anti-SIRT1 antibody produced in mouse, clone 7B7, purified immunoglobulin, buffered aqueous solution
SAB1404972
Monoclonal Anti-SIRT1, (C-terminal) antibody produced in mouse, clone 7C2, purified immunoglobulin, buffered aqueous solution
PLA0132
Rabbit anti-Sirt1 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
S8446 SIRT1 human, recombinant, expressed in E. coli, N-terminal histidine tagged, ≥90% (SDS-PAGE), buffered aqueous glycerol solution
SRP0115
Sirtuin 1 human, recombinant, expressed in E. coli, ≥60% (SDS-PAGE)