EMAIL THIS PAGE TO A FRIEND
Biochemistry
Cancer-Associated Mutations in Breast Tumor Kinase/PTK6 Differentially Affect Enzyme Activity and Substrate Recognition.
PMID 25940761
Abstract
Brk (breast tumor kinase, also known as PTK6) is a nonreceptor tyrosine kinase that is aberrantly expressed in several cancers and promotes cell proliferation and transformation. Genome sequencing studies have revealed a number of cancer-associated somatic mutations in the Brk gene; however, their effect on Brk activity has not been examined. We analyzed a panel of cancer-associated mutations and determined that several of the mutations activate Brk, while two eliminated enzymatic activity. Three of the mutations (L16F, R131L, and P450L) are located in important regulatory domains of Brk (the SH3, SH2 domains, and C-terminal tail, respectively). Biochemical data suggest that they activate Brk by disrupting intramolecular interactions that normally maintain Brk in an autoinhibited conformation. We also observed differential effects on recognition and phosphorylation of substrates, suggesting that the mutations can influence downstream Brk signaling by multiple mechanisms.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1406846
Anti-BCAR1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1401483
Anti-BCAR1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA042282
Anti-BCAR1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2502079
Anti-ESR2 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
AV31652
Anti-ESR2 antibody produced in rabbit, affinity isolated antibody
SAB2100713
Anti-ESR2 antibody produced in rabbit, affinity isolated antibody
SAB2105299
Anti-ESR2, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB1407036
Anti-KHDRBS1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV40665
Anti-KHDRBS1 antibody produced in rabbit, affinity isolated antibody
AV37130
Anti-KHDRBS1 antibody produced in rabbit, IgG fraction of antiserum
SAB2104552
Anti-KHDRBS1 antibody produced in rabbit, affinity isolated antibody
SAB4504048
Anti-phospho-p130 Cas (pTyr165) antibody produced in rabbit, affinity isolated antibody
SAB4504049
Anti-phospho-p130 Cas (pTyr249) antibody produced in rabbit, affinity isolated antibody
SAB4503824
Anti-phospho-p130 Cas (pTyr410) antibody produced in rabbit, affinity isolated antibody
S9575
Anti-Sam68 (C-terminal) antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution
S9450
Anti-Sam68 (N-terminal) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
WH0002100M1
Monoclonal Anti-ESR2 antibody produced in mouse, clone 3F3, purified immunoglobulin, buffered aqueous solution
SAB2702145
Monoclonal Anti-ESR2 antibody produced in mouse
SAB2702146 Monoclonal Anti-ESR2 antibody produced in mouse
SAB2702147 Monoclonal Anti-ESR2 antibody produced in mouse
E1276 Monoclonal Anti-Estrogen Receptor β antibody produced in mouse, clone 9.88, ascites fluid
WH0010657M3
Monoclonal Anti-KHDRBS1 antibody produced in mouse, clone 1A4, purified immunoglobulin, buffered aqueous solution