EMAIL THIS PAGE TO A FRIEND
Biomaterials
Intracellular surface-enhanced Raman scattering probes based on TAT peptide-conjugated Au nanostars for distinguishing the differentiation of lung resident mesenchymal stem cells.
PMID 25941778
Abstract
Lung resident mesenchymal stem cells (LR-MSCs) are important regulators of pathophysiological processes including tissue repair and fibrosis, inflammation, angiogenesis and tumor formation. Therefore, increasing attention has focused on the functional differentiation of LR-MSCs. However, the distinction between the undifferentiated and differentiated LR-MSCs, which are closely related and morphologically similar, is difficult to achieve by conventional methods. In this study, by employing the TAT Peptide-conjugated Au nanostars (AuNSs) as an intracellular probe, we developed a method for the identification of LR-MSC differentiation by surface-enhanced Raman scattering (SERS) spectroscopy. SERS spectra were analyzed using principal component analysis (PCA) that allowed unambiguous distinction of subtypes and monitoring of component changes during cellular differentiation. Furthermore, to ascertain whether co-culture with alveolar epithelial type II (ATII) cells and incubation with transform growth factor (TGF)-β were involved in regulating the differentiation of LR-MSCs, we investigated the protein expression levels of epithelial markers and fibroblastic markers on LR-MSCs. Our results demonstrated that co-culture with ATII cells or incubation with TGF-β could induce the differentiation of LR-MSCs as confirmed by SERS analysis, a method that is capable of noninvasive characterization of and distinction between subtypes of LR-MSCs during differentiation. We have provided a new tool that may facilitate stem cell research.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1306569
ANTI-CYK18(C-TERMINAL) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4501665
Anti-Keratin 18, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4300551
Anti-KRT18 (Ab-33) antibody produced in rabbit, affinity isolated antibody
AV40205
Anti-KRT18 (AB1) antibody produced in rabbit, IgG fraction of antiserum
AV40206
Anti-KRT18 (AB2) antibody produced in rabbit, IgG fraction of antiserum
GW22753 Anti-KRT18 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB2701928
Anti-krt18 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4200489
Anti-Occludin (C-terminal) antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
SAB1406198
Anti-OCLN antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA005933
Anti-OCLN antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB3500301
Anti-OCLN antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2103797
Anti-OCLN, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB3300015
Monoclonal Anti-CK18 antibody produced in mouse, clone 10, purified immunoglobulin, buffered aqueous glycerol solution
SAB3300017 Monoclonal Anti-CK18 antibody produced in mouse, clone 1H10, purified immunoglobulin, buffered aqueous glycerol solution
SAB1305526
MONOCLONAL ANTI-CYK18 (C-TERMINAL) antibody produced in mouse, clone 695CT17.2.6, crude ascites, buffered aqueous solution
SAB1305562
MONOCLONAL ANTI-CYK18(C-TERMINAL) antibody produced in mouse, clone 695CT17.2.6, IgG fraction of antiserum, buffered aqueous solution
WH0003875M1
Monoclonal Anti-KRT18 antibody produced in mouse, clone 2F8, purified immunoglobulin, buffered aqueous solution
SAB1404012
Monoclonal Anti-KRT18 antibody produced in mouse, clone 1G8, purified immunoglobulin, buffered aqueous solution
SAB4700411
Monoclonal Anti-KRT18 antibody produced in mouse, clone C-04, purified immunoglobulin, buffered aqueous solution
SAB4700670 Monoclonal Anti-KRT18 antibody produced in mouse, clone DC-10, purified immunoglobulin, buffered aqueous solution
WH0004950M1
Monoclonal Anti-OCLN antibody produced in mouse, clone 1G7, purified immunoglobulin, buffered aqueous solution
AMAB90893
Monoclonal Anti-OCLN antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1608, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90890
Monoclonal Anti-OCLN antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1567, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90889
Monoclonal Anti-OCLN antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1555, purified immunoglobulin, buffered aqueous glycerol solution