EMAIL THIS PAGE TO A FRIEND
Environmental science & technology
Anaerobic Chemolithotrophic Growth of the Haloalkaliphilic Bacterium Strain MLMS-1 by Disproportionation of Monothioarsenate.
PMID 25941832
Abstract
A novel chemolithotrophic metabolism based on a mixed arsenic-sulfur species has been discovered for the anaerobic deltaproteobacterium, strain MLMS-1, a haloalkaliphile isolated from Mono Lake, California, U.S. Strain MLMS-1 is the first reported obligate arsenate-respiring chemoautotroph which grows by coupling arsenate reduction to arsenite with the oxidation of sulfide to sulfate. In that pathway the formation of a mixed arsenic-sulfur species was reported. That species was assumed to be monothioarsenite ([H2As(III)S(-II)O2](-)), formed as an intermediate by abiotic reaction of arsenite with sulfide. We now report that this species is monothioarsenate ([HAs(V)S(-II)O3](2-)) as revealed by X-ray absorption spectroscopy. Monothioarsenate forms by abiotic reaction of arsenite with zerovalent sulfur. Monothioarsenate is kinetically stable under a wide range of pH and redox conditions. However, it was metabolized rapidly by strain MLMS-1 when incubated with arsenate. Incubations using monothioarsenate confirmed that strain MLMS-1 was able to grow (μ = 0.017 h(-1)) on this substrate via a disproportionation reaction by oxidizing the thio-group-sulfur (S(-II)) to zerovalent sulfur or sulfate while concurrently reducing the central arsenic atom (As(V)) to arsenite. Monothioarsenate disproportionation could be widespread in nature beyond the already studied arsenic and sulfide rich hot springs and soda lakes where it was discovered.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
RES1427A-A7
Ammonium sulfate, PharmaGrade, NF, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production.
H8N2O4S
A4418
Ammonium sulfate, for molecular biology, ≥99.0%
H8N2O4S
A6387
Ammonium sulfate, ReagentPlus®, ≥99.0%
H8N2O4S
A3920
Ammonium sulfate, plant cell culture tested, ≥99.0%
H8N2O4S
09978
Ammonium sulfate, BioUltra, ≥99.5% (T)
H8N2O4S
A2939
Ammonium sulfate, BioXtra, ≥99.0%
H8N2O4S
204501
Ammonium sulfate, 99.999% trace metals basis
H8N2O4S
451975
Ammonium sulfate-14N2 solution, 99.99 atom % 14N, 40 wt. % in H2O
H814N2O4S
485411
Ammonium-14N2 sulfate, 99.99 atom % 14N
H814N2O4S
485306
Ammonium-14N2 sulfate solution, 40 wt. % in H2O, 99.99 atom % 14N
H814N2O4S
608602
Ammonium-14N2,sulfate-16O4, 99.99 atom % 14N, 99.99 atom % 16O
H814N216O4S
164283
Methyl trifluoromethanesulfonate, ≥98%
C2H3F3O3S
M9140
Methylene blue, certified by the Biological Stain Commission
C16H18ClN3S · xH2O
03978
Methylene Blue solution, for microscopy, concentrate according to Ehrlich, concentrated, aqueous solution
C16H18ClN3S
1808
Methylene Blue solution, 1.4 % (w/v) in 95% ethanol
C16H18ClN3S
319112
Methylene Blue solution, 0.05 wt. % in H2O
C16H18ClN3S
S7400
Sodium (meta)arsenite, ≥90%
AsNaO2
S7026
Sodium thiosulfate
Na2O3S2
563188
Sodium thiosulfate, ≥99.99% trace metals basis
Na2O3S2
HT1005
Sodium thiosulfate solution, 2 g/dL in deionized water
Na2O3S2
213292
Sulfur, 99.998% trace metals basis
S
344621
Sulfur, flakes, ≥99.99% trace metals basis
S
414980
Sulfur, powder, 99.98% trace metals basis
S
HT-S200
Sulfur, prilled, ≥99.99 trace metals basis
S