EMAIL THIS PAGE TO A FRIEND
International journal of molecular sciences
Escherichia coli Maltose-Binding Protein Induces M1 Polarity of RAW264.7 Macrophage Cells via a TLR2- and TLR4-Dependent Manner.
PMID 25941931
Abstract
Maltose-binding protein (MBP) is a critical player of the maltose/maltodextrin transport system in Escherichia coli. Our previous studies have revealed that MBP nonspecifically induces T helper type 1 (Th1) cell activation and activates peritoneal macrophages obtained from mouse. In the present study, we reported a direct stimulatory effect of MBP on RAW264.7 cells, a murine macrophage cell line. When stimulated with MBP, the production of nitric oxide (NO), IL-1β, IL-6 and IL-12p70, and the expressions of CD80, MHC class II and inducible nitric oxide synthase (iNOS) were all increased in RAW264.7 cells, indicating the activation and polarization of RAW264.7 cells into M1 macrophages induced by MBP. Further study showed that MBP stimulation upregulated the expression of TLR2 and TLR4 on RAW264.7 cells, which was accompanied by subsequent phosphorylation of IκB-α and p38 MAPK. Pretreatment with anti-TLR2 or anti-TLR4 antibodies largely inhibited the phosphorylation of IκB-α and p38 MAPK, and greatly reduced MBP-induced NO and IL-12p70 production, suggesting that the MBP-induced macrophage activation and polarization were mediated by TLR2 and TLR4 signaling pathways. The observed results were independent of lipopolysaccharide contamination. Our study provides a new insight into a mechanism by which MBP enhances immune responses and warrants the potential application of MBP as an immune adjuvant in immune therapies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GW22787 Anti-TLR2 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
PRS3135
Anti-TLR2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1412080
ANTI-TLR4 antibody produced in mouse, clone 2D11, purified immunoglobulin, buffered aqueous solution
SAB1412548
ANTI-TLR4 antibody produced in mouse, clone 4B10, purified immunoglobulin, buffered aqueous solution
SAB1412602
ANTI-TLR4 antibody produced in mouse, clone 3G12, purified immunoglobulin, buffered aqueous solution
PRS3141
Anti-TLR4 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3500765
Anti-TLR4 antibody produced in rabbit, affinity isolated antibody
WH0007097M7
Monoclonal Anti-TLR2 antibody produced in mouse, clone 1E7, purified immunoglobulin, buffered aqueous solution
SAB1404474
Monoclonal Anti-TLR2 antibody produced in mouse, clone 2G9, purified immunoglobulin, buffered aqueous solution
SAB1409745
Monoclonal Anti-TLR2 antibody produced in mouse, clone 2A3, purified immunoglobulin, buffered aqueous solution
SAB1409746 Monoclonal Anti-TLR2 antibody produced in mouse, clone 2D2, purified immunoglobulin, buffered aqueous solution
SAB1409747 Monoclonal Anti-TLR2 antibody produced in mouse, clone 3F10, purified immunoglobulin, buffered aqueous solution
SAB1409748 Monoclonal Anti-TLR2 antibody produced in mouse, clone 1E3, purified immunoglobulin, buffered aqueous solution
SAB1409749 Monoclonal Anti-TLR2 antibody produced in mouse, clone 4D6, purified immunoglobulin, buffered aqueous solution
SAB1409750 Monoclonal Anti-TLR2 antibody produced in mouse, clone 4G9, purified immunoglobulin, buffered aqueous solution
SAB1409751 Monoclonal Anti-TLR2 antibody produced in mouse, clone 1C6, purified immunoglobulin, buffered aqueous solution
SAB1409752 Monoclonal Anti-TLR2 antibody produced in mouse, clone 1B10, purified immunoglobulin, buffered aqueous solution
SAB1409753 Monoclonal Anti-TLR2 antibody produced in mouse, clone 4C4, purified immunoglobulin, buffered aqueous solution
SAB1409754 Monoclonal Anti-TLR2 antibody produced in mouse, clone 3H8, purified immunoglobulin, buffered aqueous solution
SAB1409755 Monoclonal Anti-TLR2 antibody produced in mouse, clone 4H6, purified immunoglobulin, buffered aqueous solution
SAB1404475
Monoclonal Anti-TLR4 antibody produced in mouse, clone 3B6, purified immunoglobulin, buffered aqueous solution
SAB1402375
Monoclonal Anti-TLR4 antibody produced in mouse, clone 1H7, purified immunoglobulin, buffered aqueous solution