EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
Validation of four different spectrophotometric methods for simultaneous determination of Domperidone and Ranitidine in bulk and pharmaceutical formulation.
PMID 25942082
Abstract
Four simple, specific, accurate and precise spectrophotometric methods were developed and validated for simultaneous determination of Domperidone (DP) and Ranitidine Hydrochloride (RT) in bulk powder and pharmaceutical formulation. The first method was simultaneous ratio subtraction (SRS), the second was ratio subtraction (RS) coupled with zero order spectrophotometry (D(0)), the third was first derivative of the ratio spectra ((1)DD) and the fourth method was mean centering of ratio spectra (MCR). The calibration curve is linear over the concentration range of 0.5-5 and 1-45 μg mL(-1) for DP and RT, respectively. The proposed spectrophotometric methods can analyze both drugs without any prior separation steps. The selectivity of the adopted methods was tested by analyzing synthetic mixtures of the investigated drugs, also in their pharmaceutical formulation. The suggested methods were validated according to International Conference of Harmonization (ICH) guidelines and the results revealed that; they were precise and reproducible. All the obtained results were statistically compared with those of the reported method, where there was no significant difference.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D122
Domperidone, powder, ≥98% (HPLC)
C22H24ClN5O2
R101
Ranitidine hydrochloride, solid
C13H22N4O3S · HCl