EMAIL THIS PAGE TO A FRIEND
Environmental monitoring and assessment
Trace elements in two staple cereals (rice and wheat) and associated health risk implications in Bangladesh.
PMID 25944756
Abstract
Concentrations of fourteen trace elements (Cd, As, Pb, Cr, Ni, Zn, Se, Cu, Mo, Mn, Sb, Ba, V and Ag) in the composite samples of most frequently consumed two staple foods, i.e. rice and wheat (collected from 30 different agroecological zones for the first time in Bangladesh) were measured by ICP-MS. The mean concentrations (mg/kg fresh weight) of Cd, As, Pb, Cr, Ni, Zn, Se, Cu, Mo, Mn, Sb, Ba, V and Ag were found as 0.088, 0.321, 0.713, 0.183, 0.213, 13.178, 0.0256, 1.985, 0.102, 4.654, 0.0033, 0.144, 0.081 and 0.007 and 0.011, 0.281, 0.221, 0.352, 0.145, 15.472, 0.245, 1.894, 0.209, 22.077, 0.0012, 3.712, 0.023 and 0.0013 in rice and wheat samples, respectively. Dietary risk of human health (non-carcinogenic and carcinogenic risks) was assessed by USEPA deterministic approaches. Total target hazard quotient (THQ) values for As and Pb were higher than 1, suggesting that people would experience significant health risks from consuming rice and wheat. However, the THQ of other metals were all less than 1. Also, the estimation showed that the target carcinogenic risk (TR) of As and Pb exceeded the accepted risk level of 1 × 10(-6). Moreover, concerning the nutritional requirements of essential elements for a sound health, the recommended doses for the daily intake of Mn was conveniently supplied by the studied cereals; however, Cr, Zn, Se, Cu and Mo were below the recommend daily allowances (RDAs). Thus, the carcinogenic and non-carcinogenic risk of As and Pb with lower supplementation of essential elements via staple foods for Bangladeshi people is a matter of concern.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
277932
Thallium, granular, 1-5 mm diameter, 99.99% trace metals basis
Tl
262994
Yttrium, chips, 99.9% trace rare earth metals basis
Y
451347
Yttrium, dendritic pieces, 99.9% trace metals basis
Y
693642
Yttrium, foil, thickness 1 mm, 99.8% trace rare earth metals basis
Y
261319
Yttrium, ingot, 99.9% trace rare earth metals basis
Y
261327
Yttrium, powder, −40 mesh, 99.5% trace rare earth metals basis
Y
GF09119279
Yttrium, foil, 25x25mm, thickness 0.15mm, as rolled, 99%
Y
GF77674719
Yttrium, wire reel, 0.2m, diameter 0.5mm, 99.9%
Y
GF11301398
Yttrium, rod, 50mm, diameter 6.35mm, cast, 99%
Y
GF86394156
Yttrium, wire reel, 0.2m, diameter 1.0mm, 99.9%
Y
GF91377478
Yttrium, foil, 25x25mm, thickness 0.125mm, as rolled, 99%
Y
GF87149481
Yttrium, foil, 50x50mm, thickness 0.125mm, as rolled, 99%
Y
GF03037148
Yttrium, foil, 50x50mm, thickness 0.15mm, as rolled, 99%
Y
GF55118257
Yttrium, foil, 25mm disks, thickness 0.50mm, as rolled, 99%
Y
GF33100298
Yttrium, foil, not light tested, 100x100mm, thickness 0.025mm, as rolled, 99%
Y
GF75315719
Yttrium, foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99%
Y
GF61507740
Yttrium, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99%
Y
GF40879008
Yttrium, rod, 100mm, diameter 12.5mm, cast, 99%
Y
GF12674617
Yttrium, rod, 100mm, diameter 6.35mm, cast, 99%
Y
GF84327037
Yttrium, rod, 50mm, diameter 12.5mm, cast, 99%
Y
GF68048860
Yttrium, rod, 50mm, diameter 2.0mm, cast, 99%
Y
GF68472692
Yttrium, wire reel, 0.05m, diameter 0.5mm, 99.9%
Y
GF35512725
Yttrium, wire reel, 0.05m, diameter 1.0mm, 99.9%
Y
GF95617020
Yttrium, wire reel, 0.05m, diameter 1.0mm, square section, 99.9%
Y
GF02763963
Yttrium, wire reel, 0.1m, diameter 0.5mm, 99.9%
Y
GF39091158
Yttrium, wire reel, 0.1m, diameter 1.0mm, 99.9%
Y
GF79949255
Yttrium, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99%
Y
773972
Yttrium sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.9% trace metals basis
Y