EMAIL THIS PAGE TO A FRIEND
BioMed research international
Prevalence of Helicobacter pylori vacA, cagA, and iceA Genotypes in Cuban Patients with Upper Gastrointestinal Diseases.
PMID 25945344
Abstract
Virulence factors of Helicobacter pylori can predict the development of different gastroduodenal diseases. There are scarce reports in Cuba about H. pylori isolates genotyping. The aim of the present investigation was to identify allelic variation of the virulence genes vacA, cagA, and iceA in sixty-eight patients diagnosed as H. pylori positive by culture. In seven out of 68 patients, strains from both gastric regions were obtained and considered independent. DNA was extracted from all the H. pylori strains and evaluated by PCR-genotyping. The vacA s1 allele, cagA gene, and iceA2 allele were the most prevalent (72.0%, 56.0%, and 57.3%, respectively). Alleles from m-region showed a similar frequency as s1a and s1b subtypes. The presence of multiple H. pylori genotypes in a single biopsy and two gastric region specimens were found. Significant statistical association was observed between iceA2 allele and patients with non-peptic ulcer dyspepsia (NUD) (P = 0.037) as well as virulence genotypes (s1, s1m2) and patients over 40 years old (P < 0.05). In conclusion, the results demonstrated a high prevalence of H. pylori virulent genotypes in Cuban patients over 40 years old while iceA2 alleles demonstrated a good specificity in patients with NUD.