EMAIL THIS PAGE TO A FRIEND
Oncotarget
Torin2 targets dysregulated pathways in anaplastic thyroid cancer and inhibits tumor growth and metastasis.
PMID 25945839
Abstract
Anaplastic thyroid cancer (ATC) is rare but it is one of the most lethal human malignancies with no effective therapy. There is a pressing need to identify new therapeutic agents for ATC. We performed quantitative high-throughput screening (qHTS) in ATC cell lines using a compound library of 3,282 drugs. qHTS identified 100 pan-active agents. Enrichment analysis of qHTS data showed drugs targeting mTOR were one of the most active drug categories, and Torin2 showed the highest efficacy. We found mTOR to be upregulated in ATC. Treatment of multiple ATC cell lines with Torin2 showed significant dose-dependent inhibition of cellular proliferation with caspase-dependent apoptosis and G1/S phase arrest. Torin2 inhibited cellular migration and inhibited the phosphorylation of key effectors of the mTOR-pathway (AKT, 4E-BP1 and 70S6K), as well as claspin and survivin expression, regulators of cell cycle and apoptosis. In our in vivo mouse model of metastatic ATC, Torin2 inhibited tumor growth and metastasis and significantly prolonged overall survival. Our findings suggest that Torin2 is a promising agent for ATC therapy and that it effectively targets upregulated pathways in human ATC.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB3500210
Anti-4E-BP1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4500736
Anti-4E-BP1 antibody produced in rabbit, affinity isolated antibody
SAB4500737
Anti-4E-BP1 antibody produced in rabbit, affinity isolated antibody
SAB2100092
Anti-AKT1S1 antibody produced in rabbit, affinity isolated antibody
SAB1405470
Anti-BIRC5 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA002830
Anti-BIRC5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4300474
Anti-EIF4EBP1 (Ab-36) antibody produced in rabbit, affinity isolated antibody
SAB4300475
Anti-EIF4EBP1 (Ab-45) antibody produced in rabbit, affinity isolated antibody
HPA023501
Anti-EIF4EBP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2106955
Anti-EIF4EBP1 antibody produced in rabbit, affinity isolated antibody
SAB4503847
Anti-phospho-4E-BP1 (pSer64) antibody produced in rabbit, affinity isolated antibody
SAB4504328
Anti-phospho-4E-BP1 (pThr36) antibody produced in rabbit, affinity isolated antibody
SAB4504329
Anti-phospho-4E-BP1 (pThr45) antibody produced in rabbit, affinity isolated antibody
SAB4503848
Anti-phospho-4E-BP1 (pThr69) antibody produced in rabbit, affinity isolated antibody
SAB4504686
Anti-phospho-4E-BP1 (pThr70) antibody produced in rabbit, affinity isolated antibody
SAB4300179
Anti-phospho-EIF4EBP1 (pThr45) antibody produced in rabbit, affinity isolated antibody
SAB4200407
Anti-phospho-PRAS40 (pSer183) antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
SAB4200448
Anti-Phospho-PRAS40 (pThr246) antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
SAB1301801
ANTI-POTEF (CENTER) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB4200312
Anti-PRAS40 (C-terminal) antibody produced in rabbit, ~1.5 mg/mL, affinity isolated antibody
S8191
Anti-Survivin antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB3500269
Anti-Survivin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4501460
Anti-Survivin antibody produced in rabbit, affinity isolated antibody
SAB4200256
Monoclonal Anti-AKT1S1 antibody produced in mouse, ~1.0 mg/mL, clone AKT1S1-3, purified immunoglobulin
WH0000332M1
Monoclonal Anti-BIRC5 antibody produced in mouse, clone 5B10, purified immunoglobulin, buffered aqueous solution
SAB4100131
Monoclonal Anti-BIRC5 antibody produced in mouse, clone BIRC5-A113, culture supernatant
SAB5300510 Monoclonal Anti-BIRC5 antibody produced in mouse, clone 2H5H2, ascites fluid
SAB5300511
Monoclonal Anti-BIRC5 antibody produced in mouse, clone 1H5, purified immunoglobulin, buffered aqueous solution
WH0001978M1
Monoclonal Anti-EIF4EBP1 antibody produced in mouse, clone 4F3-H2, purified immunoglobulin, buffered aqueous solution
SAB1403761
Monoclonal Anti-EIF4EBP1 antibody produced in mouse, clone 1F7, purified immunoglobulin, buffered aqueous solution