EMAIL THIS PAGE TO A FRIEND
Autophagy
SYK regulates macrophage MHC-II expression via activation of autophagy in response to oxidized LDL.
PMID 25946330
Abstract
Adaptive immunity, which plays an important role in the development of atherosclerosis, is mediated by major histocompatibility complex (MHC)-dependent antigen presentation. In atherosclerotic lesions, macrophages constitute an important class of antigen-presenting cells that activate adaptive immune responses to oxidized low-density lipoprotein (OxLDL). It has been reported that autophagy regulates adaptive immune responses by enhancing antigen presentation to MHC class II (MHC-II). In a previous study, we have demonstrated that SYK (spleen tyrosine kinase) regulates generation of reactive oxygen species (ROS) and activation of MAPK8/JNK1 in macrophages. Because ROS and MAPK8 are known to regulate autophagy, in this study we investigated the role of SYK in autophagy, MHC-II expression and adaptive immune response to OxLDL. We demonstrate that OxLDL induces autophagosome formation, MHC-II expression, and phosphorylation of SYK in macrophages. Gene knockout and pharmacological inhibitors of NOX2 and MAPK8 reduced OxLDL-induced autophagy. Using bone marrow-derived macrophages isolated from wild-type and myeloid-specific SYK knockout mice, we demonstrate that SYK regulates OxLDL-induced ROS generation, MAPK8 activation, BECN1-BCL2 dissociation, autophagosome formation and presentation of OxLDL-derived antigens to CD4(+) T cells. ldlr(-/-) syk(-/-) mice fed a high-fat diet produced lower levels of IgG to malondialdehyde (MDA)-LDL, malondialdehyde-acetaldehyde (MAA)-LDL, and OxLDL compared to ldlr(-/-) mice. These results provide new insights into the mechanisms by which SYK regulates MHC-II expression via autophagy in macrophages and may contribute to regulation of adaptive immune responses in atherosclerosis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
21888
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
C29H19NO11
87444
5-Carboxy-fluorescein diacetate N-succinimidyl ester, for fluorescence, ≥95.0% (HPLC)
C29H19NO11
AV36578
Anti-ANXA3 (AB1) antibody produced in rabbit, IgG fraction of antiserum
AV36579
Anti-ANXA3 (AB2) antibody produced in rabbit, IgG fraction of antiserum
SAB1405458
Anti-ANXA3 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA013398
Anti-ANXA3 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA013431
Anti-ANXA3 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
PRS3335
Anti-Bcl-2 (ab1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS3337
Anti-Bcl-2 (ab2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
B9804 Anti-Bcl-2 antibody, Mouse monoclonal, clone 10C4, purified from hybridoma cell culture
SAB4300339
Anti-BCL2 (Ab-56) antibody produced in rabbit, affinity isolated antibody
SAB4300340
Anti-BCL2 (Ab-70) antibody produced in rabbit, affinity isolated antibody
SAB2500154
Anti-BCL2 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
AV01005
Anti-BCL2 antibody produced in rabbit, affinity isolated antibody
PRS3611
Anti-Beclin-1 (ab1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS3613
Anti-Beclin-1 (ab2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2103299
Anti-BECN1, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB1305652
ANTI-CD11B(N-TERMINAL) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1405801
Anti-FCGR2A antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA010718
Anti-FCGR2A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1405802
Anti-FCGR2B antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA014730
Anti-FCGR2B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410376
Anti-FCGR2B antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2500547
Anti-ITGAM antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1305638 ANTI-LC3 (APG8A) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1306269
ANTI-LC3 (APG8A/B) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
L8793
Anti-LC3A antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB1408113
Anti-MAP1LC3A antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA007649
Anti-MAP1LC3A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2101425
Anti-MAP1LC3A antibody produced in rabbit, affinity isolated antibody