EMAIL THIS PAGE TO A FRIEND
Cell cycle (Georgetown, Tex.)
Transcriptome profiling identifies p53 as a key player during calreticulin deficiency: Implications in lipid accumulation.
PMID 25946468
Abstract
Calreticulin (CRT) is an endoplasmic reticulum (ER) resident calcium binding protein that is involved in several cellular activities. Transcriptome analyses in CRT knockdown HepG2 cells revealed 253 altered unique genes and subsequent in silico protein-protein interaction network and MCODE clustering identified 34 significant clusters, of which p53 occupied the central hub node in the highest node-rich cluster. Toward validation, we show that CRT knockdown leads to inhibition of p53 protein levels. Both, CRT and p53 siRNA promote hepatic lipid accumulation and this was accompanied by elevated SREBP-1c and FAS levels. p53 was identified to bind at -219xa0bp on the SREBP-1c promoter and in the presence of CRT siRNA, there was decreased occupancy of p53 on this binding element. This was associated with increased SREBP-1c promoter activity and both, mutation in this binding site or p53 over-expression antagonised the effects of CRT knockdown. We, therefore, identify a negatively regulating p53 binding site on the SREBP-1c promoter that is critical during hepatic lipid accumulation. These results were validated in mouse primary hepatocytes and toward a physiological relevance, we report that while the levels of CRT and p53 are reduced in the fatty livers of diabetic db/db mice, SREBP-1c levels are significantly elevated. Our results suggest that decreased CRT levels might be involved in the development of a fatty liver by preventing p53 occupancy on the SREBP-1c promoter and thereby facilitating SREBP-1c up-regulation and consequently, lipid accumulation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1401044
Anti-CALCR antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1401045
Anti-CALCR antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA028962
Anti-CALCR antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV30114
Anti-CALR (AB1) antibody produced in rabbit, affinity isolated antibody
AV30113
Anti-CALR (AB2) antibody produced in rabbit, IgG fraction of antiserum
HPA002242
Anti-CALR antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
GW22432F Anti-Calreticulin antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
C4606
Anti-Calreticulin antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB2501990
Anti-SLC6A8 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA008802
Anti-SLC6A8 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV42248
Anti-SLC6A8 antibody produced in rabbit, IgG fraction of antiserum
SAB2104767
Anti-SLC6A8 antibody produced in rabbit, affinity isolated antibody
AV37145
Anti-SREBF1 (AB1) antibody produced in rabbit, affinity isolated antibody
SAB1412393
ANTI-SREBF1 antibody produced in mouse, clone 4G4, purified immunoglobulin, buffered aqueous solution
SAB2102992
Anti-SREBF1, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB4502850
Anti-SREBP-1 antibody produced in rabbit, affinity isolated antibody
EMU008961 MISSION® esiRNA, esiRNA targeting mouse Slc6a8 (esiRNA1)
EHU063971 MISSION® esiRNA, esiRNA human CALCR (esiRNA1)
EHU046961 MISSION® esiRNA, esiRNA human CALR (esiRNA1)
EMU016191 MISSION® esiRNA, esiRNA targeting mouse Calr (esiRNA1)
SAB1402135
Monoclonal Anti-CALCR, (C-terminal) antibody produced in mouse, clone 2F7, purified immunoglobulin, buffered aqueous solution
WH0000811M1
Monoclonal Anti-CALR antibody produced in mouse, clone 1G11-1A9, purified immunoglobulin, buffered aqueous solution
SAB5300527 Monoclonal Anti-CALR antibody produced in mouse, clone 1G6A7, ascites fluid
C7492
Monoclonal Anti-Calreticulin antibody produced in mouse, clone TO-11, tissue culture supernatant, buffered aqueous solution
WH0006720M1
Monoclonal Anti-SREBF1 antibody produced in mouse, clone 4B10, purified immunoglobulin, buffered aqueous solution
SAB1404405
Monoclonal Anti-SREBF1 antibody produced in mouse, clone 4C11, purified immunoglobulin, buffered aqueous solution