EMAIL THIS PAGE TO A FRIEND
Biological & pharmaceutical bulletin
Clostridium perfringens TpeL Induces Formation of Stress Fibers via Activation of RhoA-ROCK Signaling Pathway.
PMID 25947919
Abstract
Clostridium perfringens TpeL belongs to a family of large clostridial glucosylating cytotoxins. TpeL modifies Rac1 and Ras subfamily proteins. Herein we report TpeL-induced formation of stress fibers via RhoA-Rho kinase (ROCK) signaling. A recombinant protein (TpeL1-525) derived from the TpeL N-terminal catalytic domain in the presence of streptolysin O (SLO) induced the formation of actin stress fibers in Madin-Darby canine kidney (MDCK) cells in a dose-dependent manner. The RhoA/ROCK pathway is known to control the formation of stress fibers. We examined the role of the RhoA/ROCK pathway in TpeL-induced formation of stress fibers. TpeL1-525-induced formation of stress fibers was inhibited by the ROCK inhibitor, Y27632 and Rho protein inhibitor, C3 transferase. TpeL1-525 activated RhoA and ROCK in a dose-dependent manner. C3 transferase blocked TpeL1-525-induced activation of RhoA and ROCK whereas Y27632 inhibited TpeL-induced activation of ROCK. These results demonstrate for the first time that TpeL induces the formation of stress fibers by activating the RhoA/ROCK signaling pathway.