EMAIL THIS PAGE TO A FRIEND
Cell death & disease
Acetylation of HDAC1 and degradation of SIRT1 form a positive feedback loop to regulate p53 acetylation during heat-shock stress.
PMID 25950477
Abstract
The tumor suppressor p53 is an essential transcription factor that sensitively regulates cellular responses to various stresses. Acetylation, a critically important posttranslational modification of p53, is induced in response to cellular stresses. P53 acetylation level strongly correlates with protein stability and activity. The steady-state level of p53 acetylation is balanced by dynamic acetylation and deacetylation. Despite the function of p53 acetylation being well studied, how the steady state of p53 acetylation level is regulated in response to cellular stresses remains unclear. In particular, the dynamic regulation of the deacetylase activities responsible for p53 deacetylation during cellular stress is unknown. In the current study, we investigated the dynamic regulation of HDAC1 (histone deacetylase 1) and SIRT1 (sirtuin 1), two major enzymes for p53 deacetylation, during cell stress. We found that various cell stress events induce HDAC1 acetylation. The increased level of HDAC1 acetylation correlates with the level of p53 acetylation. Acetylated HDAC1 loses the ability to deacetylate p53. Cellular stresses also promote the decline of the SIRT1 protein in a proteasome-dependent pathway, which also results in the increase of p53 acetylation. Importantly, the decreased level of SIRT1 also contributes to the accumulation of HDAC1 acetylation as SIRT1 deacetylates HDAC1. Therefore, the increase of HDAC1 acetylation and reduced level of SIRT1 protein during cellular stress directly link to the induction of p53 acetylation. These results unveil the mechanism underlying the dynamic regulation of p53 acetylation during cell stress.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
660124
4,4′-Bis(N-carbazolyl)-1,1′-biphenyl, 97%
C36H24N2
699195
4,4′-Bis(N-carbazolyl)-1,1′-biphenyl, sublimed grade, 99.9% trace metals basis
C36H24N2
HPA003128
Anti-EP300 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA004112
Anti-EP300 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2501930
Anti-HDAC1 (C-terminal) antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2502051
Anti-Hdac1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2701686
Anti-hdac1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701822
Anti-hdac1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
AV38530
Anti-HDAC1 antibody produced in rabbit, IgG fraction of antiserum
SAB2101020
Anti-HDAC1 antibody produced in rabbit, affinity isolated antibody
HPA029693
Anti-HDAC1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4501382
Anti-HDAC1, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4501383
Anti-HDAC1, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4503485
Anti-p300, C-Terminal antibody produced in rabbit, affinity isolated antibody
P7493
Anti-PCAF antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
AV32466
Anti-PCAF antibody produced in rabbit, IgG fraction of antiserum
SAB2101734
Anti-PCAF antibody produced in rabbit, affinity isolated antibody
EMU000411 MISSION® esiRNA, esiRNA targeting mouse Sirt1 (esiRNA1)
WH0002033M2
Monoclonal Anti-EP300 antibody produced in mouse, clone 1D2, purified immunoglobulin, buffered aqueous solution
SAB1400094
Monoclonal Anti-EP300 antibody produced in mouse, clone 1B1, purified immunoglobulin, buffered aqueous solution
SAB1403776
Monoclonal Anti-EP300 antibody produced in mouse, clone 1D2, ascites fluid
WH0008850M2
Monoclonal Anti-PCAF antibody produced in mouse, clone 1H2, purified immunoglobulin, buffered aqueous solution
SAB1404637
Monoclonal Anti-PCAF antibody produced in mouse, clone 5E11, purified immunoglobulin, buffered aqueous solution
SAB1409953
Monoclonal Anti-PCAF antibody produced in mouse, clone 5E5, purified immunoglobulin, buffered aqueous solution
SAB1409954 Monoclonal Anti-PCAF antibody produced in mouse, clone 4C10, purified immunoglobulin, buffered aqueous solution
SAB1409955 Monoclonal Anti-PCAF antibody produced in mouse, clone 4D11, purified immunoglobulin, buffered aqueous solution
PLA0134
Rabbit anti-HDAC1 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
PLA0096
Rabbit anti-p300 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.