EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Qualitative and Quantitative Analysis of Lignan Constituents in Caulis Trachelospermi by HPLC-QTOF-MS and HPLC-UV.
PMID 25951006
Abstract
A high-performance liquid chromatography coupled with quadrupole tandem time-of-flight mass (HPLC-QTOF-MS) and ultraviolet spectrometry (HPLC-UV) was established for simultaneous qualitative and quantitative analysis of the major chemical constituents in Caulis Trachelospermi, respectively. The analysis was performed on an Agilent Zorbax Eclipse Plus C18 column (4.6 mm×150 mm, 5 μm) using a binary gradient system of water and methanol, with ultraviolet absorption at 230 nm. Based on high-resolution ESI-MS/MS fragmentation behaviors of the reference standards, the characteristic cleavage patterns of lignano-9, 9'-lactones and lignano-8'-hydroxy-9, 9'-lactones were obtained. The results demonstrated that the characteristic fragmentation patterns are valuable for identifying and differentiating lignano-9,9'-lactones and lignano-8'-hydroxy-9,9'-lactones. As such, a total of 25 compounds in Caulis Trachelospermi were unambiguously or tentatively identified via comparisons with reference standards or literature. In addition, 14 dibenzylbutyrolatone lignans were simultaneously quantified in Caulis Trachelospermi by HPLC-UV method. The method is suitable for the qualitative and quantitative analyses of dibenzylbutyrolatone lignans in Caulis Trachelospermi.