EMAIL THIS PAGE TO A FRIEND
Sensors (Basel, Switzerland)
Enhanced sensitive love wave surface acoustic wave sensor designed for immunoassay formats.
PMID 25951337
Abstract
We report a Love wave surface acoustic wave (LW-SAW) immunosensor designed for the detection of high molecular weight targets in liquid samples, amenable also for low molecular targets in surface competition assays. We implemented a label-free interaction protocol similar to other surface plasmon resonance bioassays having the advantage of requiring reduced time analysis. The fabricated LW-SAW sensor supports the detection of the target in the nanomolar range, and can be ultimately incorporated in portable devices, suitable for point-of-care testing (POCT) applications.