EMAIL THIS PAGE TO A FRIEND
Veterinary journal (London, England : 1997)
Mechanisms involved in quinolone resistance in Mycoplasma mycoides subsp. capri.
PMID 25951987
Abstract
Mycoplasma mycoides subsp. capri is a causative agent of contagious agalactia in goats. In this study, M. mycoides subsp. capri mutants were selected for resistance to fluoroquinolones (norfloxacin, enrofloxacin and ciprofloxacin) by serial passes in broth with increasing concentrations of antibiotic. Mutations conferring cross-resistance to the three fluoroquinolones were found in the quinolone resistance determining regions of the four genes encoding DNA gyrase and topoisomerase IV. Different mutations in the DNA gyrase GyrA subunit suggest a different mechanism of inhibition between norfloxacin and the other tested fluoroquinolones. The presence of an adenosine triphosphate-dependent efflux system was suggested through the use of the inhibitor orthovanadate.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R0875
Reserpine
C33H40N2O9