EMAIL THIS PAGE TO A FRIEND
The international journal of biochemistry & cell biology
Osteogenic differentiation of human MSCs: Specific occupancy of the mitochondrial DNA by NFATc1 transcription factor.
PMID 25952151
Abstract
A substantial body of evidence indicates that mitochondrial morphology and function change during osteogenic differentiation. However, molecular mechanisms linking mitochondrial dynamics with the regulation of osteoblast functions are poorly understood. Amongst the molecules that influence the decision of human mesenchymal stem cells (hMSCs) to become osteoblasts are Slug and NFATc1 transcription factors (TFs). These molecules also interfere with different mitochondria-dependent pathways in response to a variety of cellular demands. The present study investigated the recruitment of Slug and NFATc1 at the D-loop regulatory region of mitochondrial DNA (mtDNA) in osteogenic differentiated hMSCs with the aim of exploring whether Slug and NFATc1 also act as mitoTFs in the mitochondrial pool of nuclear TFs. The results demonstrate that NFATc1, but not Slug, is localized in the mitochondria. Using chromatin immunoprecipitation assay, we found that NFATc1 is recruited at mtDNA, but this occurs only when the calcification process is at its highest in osteo-induced MSC and the maximum level of differentiation is reached. Occupancy of the mtDNA by NFATc1 is associated with a decreased expression of crucial mitochondrial genes such as Cytochrome B and NADH dehydrogenase 1. This suggests that NFATc1 acts as a negative regulator of mtDNA transcription during the calcification process and interruption of aerobic energy demand. The finding of NFATc1 participation in osteogenic differentiation through its direct involvement in the regulatory machinery of mitochondria suggests a new role for this TF and adds information on communication between mitochondrial and nuclear genomes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB3700851 Anti-Rabbit IgG (Fc specific), highly cross adsorbed-Fluorescein antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700847 Anti-Rabbit IgG (Fc specific), highly cross adsorbed-Rhodamine antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700859 Anti-Rabbit IgG (Fc specific), highly cross adsorbed-Texas Red® antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700850 Anti-Rabbit IgG (Fc specific)-Fluorescein antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700846 Anti-Rabbit IgG (Fc specific)-Rhodamine antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700858 Anti-Rabbit IgG (Fc specific)-Texas Red® antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB4600015
Anti-Rabbit IgG (H+L), CF 350 antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600021
Anti-Rabbit IgG (H+L), CF 405M antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600024
Anti-Rabbit IgG (H+L), CF 405S antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600044
Anti-Rabbit IgG (H+L), CF 488A antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600068
Anti-Rabbit IgG (H+L), CF 555 antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600084
Anti-Rabbit IgG (H+L), CF 568 antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600107
Anti-Rabbit IgG (H+L), CF 594 antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600140
Anti-Rabbit IgG (H+L), CF 633 antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600163
Anti-Rabbit IgG (H+L), CF 640R antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600184
Anti-Rabbit IgG (H+L), CF 647 antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600193
Anti-Rabbit IgG (H+L), CF 660C antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600195
Anti-Rabbit IgG (H+L), CF 660R antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600212
Anti-Rabbit IgG (H+L), CF 750 antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600223
Anti-Rabbit IgG (H+L), F(ab′)2 fragment, CF350 antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4600406
Anti-Rabbit IgG (H+L), F(ab′)2 fragment, CF350 antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4600389
Anti-Rabbit IgG (H+L), F(ab′)2 fragment, CF488A antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4600300
Anti-Rabbit IgG (H+L), F(ab′)2 fragment, CF555 antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4600391
Anti-Rabbit IgG (H+L), F(ab′)2 fragment, CF555 antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4600400
Anti-Rabbit IgG (H+L), F(ab′)2 fragment, CF568 antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4600322
Anti-Rabbit IgG (H+L), F(ab′)2 fragment, CF594 antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4600407
Anti-Rabbit IgG (H+L), F(ab′)2 fragment, CF594 antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4600404
Anti-Rabbit IgG (H+L), F(ab′)2 fragment, CF633 antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4600398
Anti-Rabbit IgG (H+L), F(ab′)2 fragment, CF640R antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4600399
Anti-Rabbit IgG (H+L), F(ab′)2 fragment, CF640R antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution