EMAIL THIS PAGE TO A FRIEND
Hepatology (Baltimore, Md.)
In autoimmune hepatitis type 1 or the autoimmune hepatitis-sclerosing cholangitis variant defective regulatory T-cell responsiveness to IL-2 results in low IL-10 production and impaired suppression.
PMID 25953611
Abstract
Defective immune regulation plays a permissive role enabling effector cells to initiate and perpetuate tissue damage, eventually resulting in autoimmune disease. Numerical and functional regulatory T-cell (Treg) impairment has been previously reported in autoimmune liver disease (AILD; including autoimmune hepatitis and autoimmune sclerosing cholangitis ASC). However, in these early reports, Tregs were phenotypically defined as CD4(+) CD25(+) or CD4(+) CD25(high) cells. In the current study, we reexamined phenotypic and functional properties of Tregs by adopting a more refined definition of these cells that also includes negativity or low level of expression of CD127. We studied 43 AILD patients and 22 healthy subjects (HSs) and found that CD4(+) CD25(+) CD127(-) Tregs were decreased in the former. This decrease was more marked in patients with active disease than in those in remission. In AILD, Treg frequencies correlated inversely with parameters of disease activity and were not affected by immunosuppressive treatment. We also document, for the first time, that, in AILD, bona-fide Tregs produce less interleukin (IL)-10 and are impaired in their ability to suppress CD4(+) CD25(-) target cell proliferation, a feature that in HSs, but not in AILDs, is dependent, at least in part, on IL-10 secretion. Decreased IL-10 production by Tregs in AILD is linked to poor responsiveness to IL-2 and phospho signal transducer and activator of transcription 5 up-regulation. Tregs are numerically impaired in AILD, this impairment being more prominent during active disease. Notably, defective IL-10 production, resulting from low Treg responsiveness to IL-2, contributes to Treg functional impairment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2702075
Anti-AP3B1 (N-terminal) antibody produced in rabbit
SAB1409916
Anti-AP3B1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA038737 Anti-AP3B1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2106308
Anti-AP3B1 antibody produced in rabbit, affinity isolated antibody
HPA004252
Anti-CD4 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA004472
Anti-CD4 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4503583
Anti-CD4, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB2700362
Anti-Foxp3 (C-terminal) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1407427
Anti-FOXP3 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1410887
Anti-FOXP3 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2501136
Anti-GATA3 (N-terminal) antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1405853
Anti-GATA3 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV32548
Anti-GATA3 antibody produced in rabbit, affinity isolated antibody
SAB2100898
Anti-GATA3 antibody produced in rabbit, affinity isolated antibody
SAB3500250
Anti-GATA3 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA029731
Anti-GATA3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4501126
Anti-GATA3 antibody produced in rabbit, affinity isolated antibody
PRS4877
Anti-IL-17 (ab1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS4887
Anti-IL-17 (ab2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1409249
Anti-IL10 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1410712
Anti-IL10 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2502114
Anti-IL17A antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2502115
Anti-IL17A antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA052258
Anti-IL17A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2502005
Anti-IL7R antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1406026
Anti-IL7R antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1410710
Anti-IL7R antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
I5020 Anti-Interleukin 10 antibody produced in goat, IgG fraction of antiserum, lyophilized powder
GW21031 Anti-Interleukin-10 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
I5145 Anti-Interleukin-10 antibody produced in goat, IgG fraction of antiserum, lyophilized powder